Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
14514-1-AP is a rabbit polyclonal antibody raised against the N-terminal (1-247 aa) of human CREB3L2.
Immunogen sequence: MEVLESGEQ GVLQWDRKLS ELSEPGDGEA LMYHTHFSEL LDEFSQNVLG QLLNDPFLSE KSVSMEVEPS PTSPAPLIQA EHSYSLCEEP RAQSPFTHIT SDSFNDDEVE SEKWYLSTDF PSTSIKTEPV TDEPPPGLVP SVTLTITAIS TPLEKEEPPL EMNTGVDSSC QTIIPKIKLE PHEVDQFLNF SPKEGLSALP VSLWVMDMVS GSTEREYGER AGMSLYHRCC SWLYEIALFL KNKNFASK (1-247 aa encoded by BC063666)
CREB3L2 is a member of the old astrocyte specifically induced substance (OASIS) DNA binding and basic leucine zipper dimerization (bZIP) family of transcription factors, which includes CREB3 (MIM 606443) and CREB4 (MIM 607138).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-ZIB transcription factor; basic transcription factor 2; BBF2 human homolog on chromosome 7; cAMP responsive element binding protein 3-like 2; cAMP-responsive element-binding protein 3-like protein 2; Cyclic AMP-responsive element-binding protein 3-like protein 2; FUS/BBF2H7 protein; SCI-related protein Ra69; TCAG_1951439
Gene Aliases: BBF2H7; C530025K05Rik; CREB3L2; Ra69; SCIRR69
UniProt ID: (Human) Q70SY1, (Mouse) Q8BH52, (Rat) Q6QDP7
Entrez Gene ID: (Human) 64764, (Mouse) 208647, (Rat) 362339
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support