Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSQAAKASA SATVAVNPGP DTKGKGAPPA GTSPSPGTTL APTTVPITSA KAAELPPGNY RLVVFELENF QGRRAEFSGE CSNLADRGFD RVRSIIVSAG PWVAFEQSNF RGEMFILEKG EYPRWNTWSS SYRSDRLMSF RPIKMDAQEH KISLFEGANF KGNTIEIQGD DAPSLWVYGF SDRVGSVKVS SGTWVGYQYP GYRGYQYLLE PGDFRHWNEW GAFQPQMQSL RRLRDKQWHL EGSFPVLATE PPK (1-252 aa encoded by BC036790)
Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and then retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into alpha, beta, and gamma families; beta and gamma crystallins are also considered as a superfamily. Alpha and beta families are further divided into acidic and basic groups. Seven protein regions exist in crystallins: four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Beta-crystallins, the most heterogeneous, differ by the presence of the C-terminal extension (present in the basic group, none in the acidic group). Beta-crystallins form aggregates of different sizes and are able to self-associate to form dimers or to form heterodimers with other beta-crystallins. This gene, a beta basic group member, undergoes extensive cleavage at its N-terminal extension during lens maturation. It is also a member of a gene cluster with beta-A4, beta-B2, and beta-B3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-B1 crystallin; Beta-crystallin B1; crystallin, beta B1; eye lens structural protein
Gene Aliases: 3110006K12Rik; BB1CRY; CATCN3; CRYB1; CRYB11; CRYBB1; CTRCT17
UniProt ID: (Human) P53674, (Mouse) Q9WVJ5, (Rat) P02523
Entrez Gene ID: (Human) 1414, (Mouse) 12960, (Rat) 25421
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support