Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CSDE1 Antibodies

          Proteintech

          CSDE1 Polyclonal Antibody

          4 References
          View all (14) CSDE1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CSDE1 Polyclonal Antibody

          • Antibody Testing Data (11)
          CSDE1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CSDE1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CSDE1 Antibody (13319-1-AP) in ICC/IF

          Immunofluorescent analysis of (-20°C Ethanol) fixed HeLa cells using CSDE1 antibody (Product # 13319-1-AP) at dilution of 1:200 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L), CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CSDE1 Antibody in Immunocytochemistry (ICC/IF)
          CSDE1 Antibody in Immunocytochemistry (ICC/IF)
          CSDE1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSDE1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSDE1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSDE1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSDE1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSDE1 Antibody in Western Blot (WB)
          CSDE1 Antibody in Western Blot (WB)
          CSDE1 Antibody in Western Blot (WB)
          CSDE1 Antibody in Western Blot (WB)

          Product Details

          13319-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CSDE1 Fusion Protein Ag4173 (440-798 aa encoded by BC032446)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.15 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: EKEAEDGII AYDDCGVKLT IAFQAKDVEG STSPQIGDKV EFSISDKQRP GQQVATCVRL LGRNSNSKRL LGYVATLKDN FGFIETANHD KEIFFHYSEF SGDVDSLELG DMVEYSLSKG KGNKVSAEKV NKTHSVNGIT EEADPTIYSG KVIRPLRSVD PTQTEYQGMI EIVEEGDMKG EVYPFGIVGM ANKGDCLQKG ESVKFQLCVL GQNAQTMAYN ITPLRRATVE CVKDQFGFIN YEVGDSKKLF FHVKEVQDGI ELQAGDEVEF SVILNQRTGK CSACNVWRVC EGPKAVAAPR PDRLVNRLKN ITLDDASAPR LMVLRQPRGP DNSMGFGAER KIRQAGVID (440-798 aa encoded by BC032446)

          Target Information

          CSDE1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: cold shock domain containing E1, RNA binding; cold shock domain containing E1, RNA-binding; Cold shock domain-containing protein E1; N-ras upstream gene protein; NRAS-related; Protein UNR; RP5-1000E10.3; upstream of NRAS

          View more View less

          Gene Aliases: AA960392; BC016898; CSDE1; D1S155E; D3Jfr1; KIAA0885; mKIAA0885; NRU; UNR

          View more View less

          UniProt ID: (Human) O75534, (Mouse) Q91W50

          View more View less

          Entrez Gene ID: (Human) 7812, (Mouse) 229663

          View more View less

          Function(s)
          DNA binding protein binding poly(A) RNA binding nucleic acid binding RNA binding
          Process(es)
          regulation of transcription, DNA-templated nuclear-transcribed mRNA catabolic process, no-go decay male gonad development
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0