Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CSK Antibodies

          Proteintech

          CSK Polyclonal Antibody

          4 References
          View all (31) CSK antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CSK Polyclonal Antibody

          • Antibody Testing Data (23)
          CSK Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CSK Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 23

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CSK Antibody (17720-1-AP) in ICC/IF

          Immunofluorescent analysis of SH-SY5Y cells using 17720-1-AP (CSK antibody) at dilution of 1:25 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CSK Antibody in Immunocytochemistry (ICC/IF)
          CSK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CSK Antibody in Western Blot (WB)
          CSK Antibody in Western Blot (WB)
          CSK Antibody in Western Blot (WB)
          CSK Antibody in Western Blot (WB)
          CSK Antibody in Western Blot (WB)
          CSK Antibody in Immunoprecipitation (IP)

          Product Details

          17720-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:6,000
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CSK Fusion Protein Ag12103 (83-450 aa encoded by BC106073)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.32 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: FHGKITREQ AERLLYPPET GLFLVRESTN YPGDYTLCVS CDGKVEHYRI MYHASKLSID EEVYFENLMQ LVEHYTSDAD GLCTRLIKPK VMEGTVAAQD EFYRSGWALN MKELKLLQTI GKGEFGDVML GDYRGNKVAV KCIKNDATAQ AFLAEASVMT QLRHSNLVQL LGVIVEEKGG LYIVTEYMAK GSLVDYLRSR GRSVLGGDCL LKFSLDVCEA MEYLEGNNFV HRDLAARNVL VSEDNVAKVS DFGLTKEASS TQDTGKLPVK WTAPEALREK KFSTKSDVWS FGILLWEIYS FGRVPYPRIP LKDVVPRVEK GYKMDAPDGC PPAVYEVMKN CWHLDAAMRP SFLQLREQLE HIKTHELHL (83-450 aa encoded by BC106073)

          Target Information

          CSK is a tyrosine kinase that has been shown to downregulate the activity of SRC oncoprotein through phosphorylation of its carboxy terminus. Since cell transformation by SRC is caused by various mechanisms that interfere with this phosphorylation, the CSK gene might function as an antioncogene. CSK plays an important role in the regulation of cell growth, differentiation, migration, and immune responses. CSK phosphorylates tyrosine residues located in the C-terminal tails of Src-family kinases including LCK, SRC, HCK, FYN, LYN, CSK, and YES1. CSK is expressed ubiquitously as a protein of approximately 50 KDa in cytosol, but a fraction distributes to lipid rafts due to interaction with the lipid raft-associated protein PAG (Csk-binding protein).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: C-Src kinase; MGC117393; Protein-tyrosine kinase CYL; Tyrosine-protein kinase CSK

          View more View less

          Gene Aliases: CSK

          View more View less

          UniProt ID: (Human) P41240

          View more View less

          Entrez Gene ID: (Human) 1445

          View more View less

          Function(s)
          protein tyrosine kinase activity non-membrane spanning protein tyrosine kinase activity receptor binding protein binding ATP binding protein C-terminus binding kinase activity protein phosphatase binding protein kinase A catalytic subunit binding identical protein binding metal ion binding proline-rich region binding
          Process(es)
          regulation of cytokine production adaptive immune response protein phosphorylation transmembrane receptor protein tyrosine kinase signaling pathway epidermal growth factor receptor signaling pathway central nervous system development brain development negative regulation of cell proliferation negative regulation of low-density lipoprotein particle clearance cell migration cell differentiation T cell costimulation negative regulation of interleukin-6 production negative regulation of kinase activity adherens junction organization peptidyl-tyrosine autophosphorylation regulation of cell proliferation negative regulation of Golgi to plasma membrane protein transport positive regulation of MAP kinase activity innate immune response negative regulation of bone resorption oligodendrocyte differentiation negative regulation of phagocytosis T cell receptor signaling pathway regulation of Fc receptor mediated stimulatory signaling pathway negative regulation of ERK1 and ERK2 cascade cellular response to peptide hormone stimulus
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67f4cd694f-jv9gd:80/100.66.135.11:80.
          git-commit: 961b7666e63bcc533d6299fdd217d6a70bd96cf1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0