Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CSMD1 CUB and Sushi multiple domains 1 is a protein that in humans is encoded by the CSMD1gene. It is a potential tumour suppressor, the deletion of which may result in head and neck carcinomas behaving more aggressively. CSMD1 is part of the complement system that defends against pathogens through either the classical pathway or the alternative pathway. Located primarily in nerve growth cones, CSMD1 blocks the classical pathway of the immune system and is thought to be involved in tumor suppression, as defects in the gene encoding CSMD1 are associated with squamous cell carcinomas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CUB and sushi domain-containing protein 1; CUB and sushi multiple domains protein 1; multiple domain complement regulator 1; protein phosphatase 1, regulatory subunit 24
Gene Aliases: B930082H09; CSMD1; KIAA1890; Mdcr1; mKIAA1890; PPP1R24; UNQ5952/PRO19863
UniProt ID: (Human) Q96PZ7, (Mouse) Q923L3
Entrez Gene ID: (Human) 64478, (Rat) 364634, (Mouse) 94109
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support