Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MYIKQVIIQG FRSYRDQTIV DPFSSKHNVI VGRNGSGKSN LFYAIQFVLS DEFSHLRPEQ RLALLHEGTG PRVISAFVEI IFDNSDNRLP IDKEEVSLRR VIGAKKDQYF LDKKMVTKND VMNLLESAGF SRSNPYYIVK QGKINQMATA PDSQRLKLLR EVAGTRVYDE RKEESISLMK ETEGKREKIN ELLKYIEERL HTLEEEKEEL AQYQKWDKMR RALEYTIYNQ ELNETRAKLD ELSAKRETSG EKSRQLRDAQ QDARDKMEDI ERQVRELKTK ISAMKEEKEQ LSAERQEQIK QRTKLELKAK DLQDELAGNS EQRKRLLKER QKLLEKIEEK QKELAETEPK FNSVKEKEER GIARLAQATQ ERTDLYAKQG RGSQFTSKEE RDKWIKKELK SLDQAINDKK RQIAAIHKDL EDTEANKEKN LEQYNKLDQD LNEVKARVEE LDRKYYEVKN KKDELQSERN YLWREENAEQ QALAAKREDL EKKQQLLRAA TGKAILNGID SINKVLDHFR RKGINQHVQN GYHGIVMNNF ECEPAFYTCV EVTAGNRLFY HIVDSDEVST KILMEFNKMN LPGEVTFLPL NKLDVRDTAY PETNDAIPMI SKLRYNPRFD KAFKHVFGKT LICRSMEVST QLARAFTMDC ITLEGDQVSH RGALTGGYYD TRKSRLELQK DVRKAEEELG ELEAKLNENL RRNIERINNE IDQLMNQMQQ IETQQRKFKA SRDSILSEMK MLKEKRQQSE KTFMPKQRSL QSLEASLHAM ESTRESLKAE LGTDLLSQLS LEDQKRVDAL NDEIRQLQQE NRQLLNERIK LEGIITRVET YLNENLRKRL DQVEQELNEL RETEGGTVLT ATTSELEAIN KRVKDTMARS EDLDNSIDKT EAGIKELQKS MERWKNMEKE HMDAINHDTK ELEKMTNRQG MLLKKKEECM KKIRELGSLP QEAFEKYQTL SLKQLFRKLE QCNTELKKYS HVNKKALDQF VNFSEQKEKL IKRQEELDRG YKSIMELMNV LELRKYEAIQ LTFKQVSKNF SEVFQKLVPG GKATLVMKKG DVEGSQSQDE GEGSGESERG SGSQSSVPSV DQFTGVGIRV SFTGKQGEMR EMQQLSGGQK SLVALALIFA IQKCDPAPFY LFDEIDQALD AQHRKAVSDM IMELAVHAQF ITTTFRPELL ESADKFYGVK FRNKVSHIDV ITAEMAKDFV EDDTTHG
This gene belongs to the SMC3 subfamily of SMC proteins. The encoded protein occurs in certain cell types as either an intracellular, nuclear protein or a secreted protein. The nuclear form, known as structural maintenance of chromosomes 3, is a component of the multimeric cohesin complex that holds together sister chromatids during mitosis, enabling proper chromosome segregation. Post-translational modification of the encoded protein by the addition of chondroitin sulfate chains gives rise to the secreted proteoglycan bamacan, an abundant basement membrane protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bamacan; Basement membrane-associated chondroitin proteoglycan; Chondroitin sulfate proteoglycan 6; chondroitin sulfate proteoglycan 6 (bamacan); Chromosome-associated polypeptide; hCAP; SMC protein 3; Structural maintenance of chromosomes protein 3
Gene Aliases: BAM; BMH; CDLS3; CSPG6; HCAP; SMC3; SMC3L1
UniProt ID: (Human) Q9UQE7
Entrez Gene ID: (Human) 9126
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support