Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MCSCYWIFC CSPFFVEKFR SVRSRLYVGR EQKLGATLSG LIEEKCKLLE KFSLIQKEYE GYEVESSLED ASFEKAAAEE ARSLEATCEK LNRSNSELED EILCLEKDLK EEKSKHSQQD ELMADISKSI QSLEDESKSL KSQIAEAKII CKTFKMSEER RAIAIKDALN ENSQLQTSHK QLFQQEAEVW KGEVSELNKQ KITFEDSKVH AEQVLNDKEN HIKTLTGHLP MMKDQAAVLE EDTTDDDNLE LEVNSQWENG ANLDDPLKGA LKKLIHAAKL NVSLKSLEGE RNHIIIQLSE VDKTKEELTE HIKNLQTQQE SLQSENIYFE SENQKLQQKL KIMTEFYQED E (1-350 aa encoded by BC043153)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cTAGE family member 6; CTAGE family, member 6; CTAGE family, member 6, pseudogene; cutaneous T-cell lymphoma-associated antigen 6; Protein cTAGE-6; putative protein cTAGE-6
Gene Aliases: CTAGE6; CTAGE6P
UniProt ID: (Human) Q86UF2
Entrez Gene ID: (Human) 340307
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support