Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTSATSPIIL KWDPKSLEIR TLTVERLLEP LVTQVTTLVN TSNKGPSGKK KGRSKKAHVL AASVEQATQN FLEKGEQIAK ESQDLKEELV AAVEDVRKQG ETMRIASSEF ADDPCSSVKR GTMVRAARAL LSAVTRLLIL ADMADVMRLL SHLKIVEEAL EAVKNATNEQ DLANRFKEFG KEMVKLNYVA ARRQQELKDP HCRDEMAAAR GALKKNATML YTASQAFLRH PDVAATRANR DYVFKQVQEA IAGISNAAQA TSPTDEAKGH TGIGELAAAL NEFDNKIILD PMTFSEARFR PSLEERLESI ISGAALMADS SCTRDDRRER IVAECNAVRQ ALQDLLSEYM NNTGRKEKGD PLNIAIDKMT KKTRDLRRQL RKAVMDHISD SFLETNVPLL VLIEAAKSGN EKEVKEYAQV FREHANKLVE VANLACSISN NEEGVKLVRM AATQIDSLCP QVINAALTLA ARPQSKVAQD NMDVFKDQWE KQVRVLTEAV DDITSVDDFL SVSENHILED VNKCVIALQE GDVDTLDRTA GAIRGRAARV IHIINAEMEN YEAGVYTEKV LEATKLLSET VMPRFAEQVE VAIEALSANV PQPFEENEFI DASRLVYDGV RDIRKAVLMI RTPEELEDDS DFEQEDYDVR SRTSVQTEDD QLIAGQSARA IMAQLPQEEK AKIAEQVEIF HQEKSKLDAE VAKWDDSGND IIVLAKQMCM IMMEMTDFTR GKGPLKNTSD VINAAKKIAE AGSRMDKLAR AVADQCPDSA CKQDLLAYLQ RIALYCHQLN ICSKVKAEVQ NLGGELIVSG LDSATSLIQA AKNLMNAVVL TVKASYVAST KYQKVYGTAA VNSPVVSWKM KAPEKKPLVK REKPEEFQTR VRRGSQKKHI SPVQALSEFK AMDSF
Alpha catenin is a member of the b-catenin family of cytosolic proteins. Alpha catenin interacts with N-cadherin and b-catenin on the cytosolic side of adherins junctions to mediate the interaction with the actin cytoskeleton. Alpha catenin is a homodimer in solution, but dissociates to form a 1:1 heterodimer with b-catenin in order to interact with cadherins. It also regulates morphological plasticity of synapses and cerebellar and hippocampal lamination during development. Alpha catenin also functions in the control of the startle modulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha E-catenin; Alpha N-catenin; alpha-catenin; Alpha-catenin-related protein; alpha-E-catenin; alpha-N-catenin; alphaE-catenin; alphaN-catenin; Cadherin-associated protein; cadherin-associated protein, related; cancer/testis antigen 114; catenin; catenin (cadherin associated protein), alpha 1; catenin (cadherin-associated protein), alpha 1, 102kDa; catenin (cadherin-associated protein), alpha 2; catenin alpha 2; Catenin alpha-1; Catenin alpha-2; Catenin alpha1; Catenin alpha2; catenin, alpha 1; Catenin-alpha1; Catenin-alpha2; FLJ36832; FLJ52416; HGNC:2509; NY-REN-13 antigen; Renal carcinoma antigen NY-REN-13
Gene Aliases: CAP-R; CAP102; CAPR; Catna1; Catna2; CT114; CTNNA1; CTNNA2; CTNR; MDPT2
UniProt ID: (Human) P35221, (Human) P26232
Entrez Gene ID: (Human) 1495, (Human) 1496, (Rat) 307505, (Rat) 297357
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support