Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence homology: Cow: 93%; Dog: 86%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 79%; Rat: 93%
Preferential cleavage: Tyr- -Xaa, Trp- -Xaa, Phe- -Xaa, Leu- -Xaa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chymotrypsinogen B
Gene Aliases: CTRB; CTRB1
UniProt ID: (Human) P17538
Entrez Gene ID: (Human) 1504
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support