Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNTQKGSLT INVHRGSLAM SIQRGSLVPR DMDSSGRDMQ LRVIPAEVKF LDTMAGRVYR LPITVHNICR WNQKIRFKEP VKPQFKLMLT SLDKELASGL QMTAMVEYHP DKDEDTFDRL LISIENKTTE IPLIGLIPSC QLEIESVVNF GTLVANSKVY SKEITITNHG KAPGIFKAEY HGQLPILIFP TSGIVDAKSS MVIKVDFCAD QPRIVDEEAI VILQGQPEML LSIKAHVVEQ IIELLSMSSD RRLECIHFGP VFFGSSKIKH ARVYNNSPEP INWVAIIQDD AVGEELGTDI QQRTDIALNN LTYIRKIKNI DTTIIISCLP NEGTLQPYQK TVITFCFTPK LM (1-351 aa encoded by BC027936)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calponin homology domain-containing protein 2; Cilia- and flagella-associated protein 47
Gene Aliases: CFAP47; CHDC2; CXorf22; CXorf30; CXorf59
UniProt ID: (Human) Q6ZTR5
Entrez Gene ID: (Human) 286464
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support