Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: deleted in autism-1 related protein; Deleted in autism-related protein 1; Divergent protein kinase domain 2B; hCG1981635; UPF0672 protein CXorf36
Gene Aliases: 4930578C19Rik; bA435K1.1; CXorf36; DIA1R; DIPK2B; EPQL1862; PRO3743; UNQ1862/PRO3743
UniProt ID: (Human) Q9H7Y0
Entrez Gene ID: (Human) 79742
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support