Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: NT2D1, IMR32, U87-MG.
Predicted reactivity: Rat (80%), Rhesus Monkey (99%), Chimpanzee (99%), Bovine (84%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NCB5OR is a flavohemoprotein that contains functional domains found in both cytochrome b5 (CYB5A; MIM 250790) and CYB5 reductase (see MIM 250800).
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: b5+b5R; cb5/cb5R; cytochrome b-type NAD(P)H oxidoreductase; Cytochrome b5 reductase 4; flavohemoprotein b5+b5R; Flavohemoprotein b5/b5R; N-terminal cytochrome b5 and cytochrome b5 oxidoreductase domain-containing protein; NADPH cytochrome B5 oxidoreductase; RP4-676J13.1
Gene Aliases: cb5/cb5R; CYB5R4; dJ676J13.1; NCB5OR
UniProt ID: (Human) Q7L1T6
Entrez Gene ID: (Human) 51167
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support