Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MALRAKAEVC MAVPWLSLQR AQALGTRAAR VPRTVLPFEA MPRRPGNRWL RLLQIWREQG YEDLHLEVHQ TFQELGPIFR SRHSASFGRW GRSAARAGLW RCQGRGWCRA NPSSLQRGQD SEALKYDLGG AGMVCVMLPE DVEKLQQVDS LHPHRMSLEP WVAYRQHRGH KCGVFLLNVA DRGNSSPPFP GGIHGAPTHS GCRNGPEWRF NRLRLNPEVL SPNAVQRFLP MVDAVARDFS QALKKKVLQN ARGSLTLDVQ PSIFHYTIEA SNLALFGERL GLVGHSPSSA SLNFLHALEV MFKSTVQLMF MPRSLSRWTS PKVWKEHFEA WDCIFQYGDN CIQKIYQELA FSRPQQYTSI VAELLLNAEL SPDAIKANSM ELTAGSVDTT VFPLLMTLFE LARNPNVQQA LRQESLAAAA SISEHPQKAT TELPLLRAAL KETLRLYPVG LFLERVASSD LVLQNYHIPA GTLVRVFLYS LGRNPALFPR PERYNPQRWL DIRGSGRNFY HVPFGFGMRQ CLGRRLAEAE MLLLLHHVLK HLQVETLTQE DIKMVYSFIL RPSMFPLLTF RAIN
The steroid 11beta-hydroxylase gene, also designated Cyp11b-1, is a marker for the functional differentiation of cells in the zonae fasciculata reticularis. The deduced protein CYP11B1 consists of 466 amino acids containing a secretory signal, epidermal growth factor-like repeats, and a proteolytically inactive cathepsin B-related sequence. A related protein, human aldosterone synthase (CYP11B2), is involved in substrate recognition and conversion, with a functionally significant residue 112 in the N-terminal region of human CYP11B2. The inherited disorder glucocorticoid-remediable aldosteronism is caused by a chimeric gene duplication between the CYP11B1 and CYP11B2 genes. This disorder is characterized by hyperaldosteronism and high levels of 18-hydroxycortisol and 18-oxocortisol, which are under ACTH control.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CYP11B1; CYPXIB1; Cytochrome P-450c11; Cytochrome P450 11B1, mitochondrial; cytochrome p450 XIB1; cytochrome P450, family 11, subfamily B, polypeptide 1; cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 1; Cytochrome P450C11; DKFZp686B05283; FLJ36771; P450C 11; steroid 11-beta-hydroxylase; Steroid 11-beta-hydroxylase, CYP11B1; steroid 11-beta-monooxygenase
Gene Aliases: CPN1; CYP11B; CYP11B1; FHI; P450C11; S11BH
UniProt ID: (Human) P15538
Entrez Gene ID: (Human) 1584
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support