Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLDFAIFAVT FLLALVGAVL YLYPASRQAA GIPGITPTEE KDGNLPDIVN SGSLHEFLVN LHERYGPVVS FWFGRRLVVS LGTVDVLKQH INPNKTLDPF ETMLKSLLRY QSGGGSVSEN HMREKLYENG VTDSLKSNFA LLLKLSEELL DKWLSYPETQ HVPLSQHMLG FAMKSVTQMV MGSTFEDDQE VIRFQKNHGT VWSEIGKGFL DGSLDKNMTR KKQYEDALMQ LESVLRNIIK ERKGRNFSQH IFIDSLVQGN LNDQQILEDS MIFSLASCII TAKLCTWAIC FLTTSEEVQK KLYEEINQVF GNGPVTPEKI EQLRYCQHVL CETVRTAKLT PVSAQFQDIE GKIDRFIIPR ETLVLYALGV VLQDPNTWPS PHKFDPDRFD DELVMKTFSS LGFSGTQECP ELRFAYMVTT VLLSVLVKRL HLLSVEGQVI ETKYELVTSS REEAWITVSK RY
CYP20A1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. The protein lacks one amino acid of the conserved heme binding site. It also lacks the conserved I-helix motif AGX(D, E)T, suggesting that its substrate may carry its own oxygen.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytochrome P450 20A1; cytochrome P450 monooxygenase; cytochrome P450, family 20, subfamily A, polypeptide 1
Gene Aliases: CYP-M; CYP20A1; UNQ667/PRO1301
UniProt ID: (Human) Q6UW02
Entrez Gene ID: (Human) 57404
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support