Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CYP20A1 Antibodies

          Invitrogen

          CYP20A1 Polyclonal Antibody

          View all (9) CYP20A1 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CYP20A1 Polyclonal Antibody

          • Antibody Testing Data (1)
          CYP20A1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CYP20A1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CYP20A1 Antibody (PA5-109611) in WB

          Western blot analysis of CYP20A1 in extracts of various cell lines. Samples were incubated with CYP20A1 Polyclonal antibody (Product # PA5-109611) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Enhanced Kit. Exposure time: 90s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CYP20A1 Antibody in Western Blot (WB)

          Product Details

          PA5-109611

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 183-462 of human CYP20A1 (NP_803882.1).
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.63 mg/mL

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          RRID

          AB_2855022

          Product Specific Information

          Immunogen sequence: STFEDDQEVI RFQKNHGTVW SEIGKGFLDG SLDKNMTRKK QYEDALMQLE SVLRNIIKER KGRNFSQHIF IDSLVQGNLN DQQILEDSMI FSLASCIITA KLCTWAICFL TTSEEVQKKL YEEINQVFGN GPVTPEKIEQ LRYCQHVLCE TVRTAKLTPV SAQLQDIEGK IDRFIIPRET LVLYALGVVL QDPNTWPSPH KFDPDRFDDE LVMKTFSSLG FSGTQECPEL RFAYMVTTVL LSVLVKRLHL LSVEGQVIET KYELVTSSRE EAWITVSKRY

          Target Information

          CYP20A1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. The protein lacks one amino acid of the conserved heme binding site. It also lacks the conserved I-helix motif AGX(D, E)T, suggesting that its substrate may carry its own oxygen.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cytochrome P450 20A1; cytochrome P450 monooxygenase; cytochrome P450, 20a1; cytochrome P450, family 20, subfamily A, polypeptide 1

          View more View less

          Gene Aliases: A930011N14Rik; CYP-M; CYP20A1; Cypm; UNQ667/PRO1301

          View more View less

          UniProt ID: (Human) Q6UW02, (Mouse) Q8BKE6, (Rat) Q6P7D4

          View more View less

          Entrez Gene ID: (Human) 57404, (Mouse) 77951, (Rat) 316435

          View more View less

          Function(s)
          monooxygenase activity iron ion binding oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen heme binding oxidoreductase activity fluorene oxygenase activity mono-butyltin dioxygenase activity tri-n-butyltin dioxygenase activity di-n-butyltin dioxygenase activity methylsilanetriol hydroxylase activity methyl tertiary butyl ether 3-monooxygenase activity 4-nitrocatechol 4-monooxygenase activity 4-chlorophenoxyacetate monooxygenase activity tert-butanol 2-monooxygenase activity alpha-pinene monooxygenase activity dimethylsilanediol hydroxylase activity ammonia monooxygenase activity hydroxymethylsilanetriol oxidase activity 2-hydroxyisobutyrate 3-monooxygenase activity alpha-pinene dehydrogenase activity bisphenol A hydroxylase B activity 2,2-bis(4-hydroxyphenyl)-1-propanol hydroxylase activity 9-fluorenone-3,4-dioxygenase activity anthracene 9,10-dioxygenase activity 2-(methylthio)benzothiazole monooxygenase activity 2-hydroxybenzothiazole monooxygenase activity benzothiazole monooxygenase activity 2,6-dihydroxybenzothiazole monooxygenase activity pinacolone 5-monooxygenase activity thioacetamide S-oxygenase activity thioacetamide S-oxide S-oxygenase activity endosulfan monooxygenase I activity N-nitrodimethylamine hydroxylase activity 4-(1-ethyl-1,4-dimethyl-pentyl)phenol monoxygenase activity endosulfan ether monooxygenase activity pyrene 4,5-monooxygenase activity pyrene 1,2-monooxygenase activity 1-hydroxypyrene 6,7-monooxygenase activity 1-hydroxypyrene 7,8-monooxygenase activity phenylboronic acid monooxygenase activity spheroidene monooxygenase activity metal ion binding
          Process(es)
          oxidation-reduction process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-9267x:80/100.66.134.224:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0