Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKRRKYGFIY KTHLFGRPTV RVMGADNVRR ILLGEHRLVS VHWPASVRTI LGSGCLSNLH DSSHKQRKKV IMRAFSREAL ECYVPVITEE VGSSLEQWLS CGERGLLVYP EVKRLMFRIA MRILLGCEPQ LAGDGDSEQQ LVEAFEEMTR NLFSLPIDVP FSGLYRGMKA RNLIHARIEQ NIRAKICGLR ASEAGQGCKD ALQLLIEHSW ERGERLDMQA LKQSSTELLF GGHETTASAA TSLITYLGLY PHVLQKVREE LKSKGLLCKS NQDNKLDMEI LEQLKYIGCV IKETLRLNPP VPGGFRVALK TFELNGYQIP KGWNVIYSIC DTHDVAEIFT NKEEFNPDRF MLPHPEDASR FSFIPFGGGL RSCVGKEFAK ILLKIFTVEL ARHCDWQLLN GPPTMKTSPT VYPVDNLPAR FTHFHGEI
CYP26A1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum protein acts on retinoids, including all-trans-retinoic acid (RA), with both 4-hydroxylation and 18-hydroxylation activities. This enzyme regulates the cellular level of retinoic acid which is involved in regulation of gene expression in both embryonic and adult tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CYP26A1; Cytochrome P450 26A1; Cytochrome P450 retinoic acid-inactivating 1; cytochrome P450, family 26, subfamily A, polypeptide 1; cytochrome P450, subfamily XXVIA, polypeptide 1; Cytochrome P450RAI; hP450RAI; P450, retinoic acid-inactivating, 1; Retinoic acid 4-hydroxylase; Retinoic acid-metabolizing cytochrome
Gene Aliases: CP26; CYP26; CYP26A1; P450RAI; P450RAI1
UniProt ID: (Human) O43174
Entrez Gene ID: (Human) 1592
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support