Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEPFVVLVLC LSFMLLFSLW RQSCRRRKLP PGPTPLPIIG NMLQIDVKDI CKSFTNFSKV YGPVFTVYFG MNPIVVFHGY ESVKEALIDN GEEFSGRGNS PISQRITKGL GIISSNGKRW KEIRRFSLTT LRNFGMGKRS IEDRVQEEAH CLVEELRKTK ASPCDPTFIL GCAPCNVICS VVFQKRFDYK DQNFLTLMKR FNENFRILNS PWIQVCNNFP LLIDCFPGTH NKVLKNVALT RSYIREKVKE HQASLDVNNP RDFIDCFLIK MEQEKDNQKS EFNIENLVGT VADLFVAGTE TTSTTLRYGL LLLLKHPEVT AKVQEEIDHV IGRHRSPCMQ DRSHMPYTDA VVHEIQRYSD LVPTGVPHAV TTDTKFRNYL IPKGTTIMAL LTSVLHDDKE FPNPNIFDPG HFLDKNGNFK KSDYFMPFSA GKRICAGEGL ARMELFLFLT TILQNFNLKS VDDLKNLNTT AVTKGIVSLP PSYQICFIPV
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by phenobarbital. The enzyme is known to metabolize many xenobiotics, including the anticonvulsive drug mephenytoin, benzo(a)pyrene, 7-ethyoxycoumarin, and the anti-cancer drug taxol. This gene is located within a cluster of cytochrome P450 genes on chromosome 10q24.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CYPIIC8; Cytochrome P450 2C8; Cytochrome P450 form 1; Cytochrome P450 IIC2; Cytochrome P450 MP-12; Cytochrome P450 MP-20; cytochrome P450, family 2, subfamily C, polypeptide 8; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 8; flavoprotein-linked monooxygenase; microsomal monooxygenase; P450 form 1; P450 IIC2; P450 MP-12/MP-20; S-mephenytoin 4-hydroxylase; xenobiotic monooxygenase
Gene Aliases: CPC8; CYP2C8; CYPIIC8; MP-12/MP-20
UniProt ID: (Human) P10632
Entrez Gene ID: (Human) 1558
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support