Search Thermo Fisher Scientific
Heat-mediated antigen retrieval is recommended for the staining of paraffin sections.
Mouse anti Feline calicivirus antibody, clone FCV1-43 recognizes feline calicivirus, and does not cross-react with FIV or FeLV.