Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CAPNS2 Antibodies

          Proteintech

          Calpain S2 Polyclonal Antibody

          View all (7) CAPNS2 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Calpain S2 Polyclonal Antibody

          • Antibody Testing Data (3)
          Calpain S2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Calpain S2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Calpain S2 Antibody (15803-1-AP) in WB

          Mouse skin tissue were subjected to SDS PAGE followed by western blot with 15803-1-AP (Calpain S2 antibody) at dilution of 1:300 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Calpain S2 Antibody in Western Blot (WB)
          Calpain S2 Antibody in Western Blot (WB)
          Calpain S2 Antibody in Immunoprecipitation (IP)

          Product Details

          15803-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200-1:1,000
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Calpain S2 Fusion Protein Ag8534 (5-248 aa encoded by BC005397)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.3 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: KALLEGADR GLGEALGGLF GGGGQRREGG GRNIGGIVGG IVNFISEAAA AQYTPEPPPT QQHFTSVEAS ESEEVRRFRQ QFTQLAGPDM EVGATDLMNI LNKVLSKHKD LKTDGFSLDT CRSIVSVMDS DTTGKLGFEE FKYLWNNIKK WQCVYKQYDR DHSGSLGSSQ LRGALQAAGF QLNEQLYQMI VRRYANEDGD MDFNNFISCL VRLDAMFRAF KSLDRDRDGL IQVSIKEWLQ LTMYS (5-248 aa encoded by BC005397)

          Target Information

          The Calpain family of proteins are calcium-regulated thiol proteases which have broad endopeptidase activity throughout the body. Calpain small subunit 2, also known as CSS2 or CAPNS2, is a calcium-dependent protease that is expressed ubiquitously in the cytoplasm. Part of a heterodimer composed of a small subunit and a large subunit, CSS2 catalyzes proteolysis of various proteins involved in cytoskeletal remodeling and signal transduction. CSS2 also acts as a chaperone to the larger subunit, mediating its correct folding and conformation. When bound as a heterodimer, CSS2 is thought to keep the catalytic activity of the large subunit dormant. After binding calcium, CSS2 is released from the complex, thereby activating the large subunit and allowing CSS2 to translocate from the cytoplasm to the cell membrane. Defects in the gene encoding CSS2 result in incorrect calpain activity and retarded fetal development, suggesting that CSS2 expression is essential for proper growth.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Calcium-dependent protease small subunit 2; Calpain small subunit 2; calpain, small subunit 2; CSS2

          View more View less

          Gene Aliases: 2310005G05Rik; 30K-2; CAPNS2

          View more View less

          UniProt ID: (Human) Q96L46, (Mouse) Q9D7J7

          View more View less

          Entrez Gene ID: (Human) 84290, (Mouse) 69543, (Rat) 679870

          View more View less

          Function(s)
          calcium-dependent cysteine-type endopeptidase activity calcium ion binding metal ion binding
          Process(es)
          proteolysis extracellular matrix disassembly
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fd48b9cb8-69xq6:80/100.66.134.130:80.
          git-commit: fe0b5dd8e7a67b57f27a6e2d26c18f8954d91d59
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0