{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Compatible with Direct ELISA
Camoteskimab is a monoclonal antibody that targets and inhibits the activity of interleukin-31 (IL-31), a cytokine involved in inflammatory and pruritic (itch-related) responses.
Protein Aliases: IGIF; IL18; IL1F4