Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody detects endogenous levels of total CPA4.
Carboxypeptidase A4 is a member of the carboxypeptidase A/B subfamily. Carboxypeptidases are zinc-containing exopeptidases that catalyze the release of carboxy-terminal amino acids, and are synthesized as zymogens that are activated by proteolytic cleavage. This protein could be involved in the histone hyperacetylation pathway. It is imprinted and may be a strong candidate protein for prostate cancer aggressiveness.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carboxypeptidase A3; Carboxypeptidase A4
Gene Aliases: 1110019K20Rik; AV009555; CPA3; CPA4; UNQ694/PRO1339
UniProt ID: (Human) Q9UI42, (Mouse) Q6P8K8
Entrez Gene ID: (Human) 51200, (Mouse) 71791
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support