Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Caveolin 1 Antibodies

          Rockland

          Caveolin-1 Polyclonal Antibody

          View all (47) Caveolin 1 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Caveolin-1 Polyclonal Antibody

          Product Details

          600-401-J64

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000
          -

          ELISA (ELISA)

          1: 10,000
          -

          Immunoprecipitation (IP)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Anti-Caveolin 1 affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the internal region of human Caveolin-1 protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          0.02M potassium phosphate, pH 7.2, with 50% glycerol, 0.15M NaCl

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.

          Anti-Caveolin-1 pS168 is directed against the phosphorylated form of human S168 residue. A BLAST analysis was used to suggest cross-reactivity with primate, human, horse, cat, and dog based on 100% sequence homology. Cross-reactivity with Caveolin-1 pS168 from other sources has not been determined.

          Target Information

          Caveolin-1 or CAV1 is a scaffolding proteins within caveolar membranes. It interacts directly with G-protein alpha subunits and can functionally regulate their activity. Caveolin-1 is also involved in the co-stimulatory signal essential for T-cell receptor (TCR) mediated T-cell activation. It can bind with DPP4 and induces T-cell proliferation and NF-kappa-B activation in a T-cell receptor/CD3-dependent manner. Mutations affecting the CAV1 gene can result in congenital generalized lipodystrophy 3, pulmonary hypertension primary 3, or partial lipodystrophy/congenital cataracts, and neurodegeneration syndrome.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: caveolae; caveolin 1, caveolae protein, 22kDa; Caveolin-1; cell growth-inhibiting protein 32; HGNC:1527; OTTHUMP00000196475

          View more View less

          Gene Aliases: BSCL3; CAV; CAV1; CGL3; LCCNS; MSTP085; PPH3; VIP21

          View more View less

          UniProt ID: (Human) Q03135

          View more View less

          Entrez Gene ID: (Human) 857

          View more View less

          Function(s)
          RNA polymerase I transcription factor binding receptor binding patched binding structural molecule activity protein binding cholesterol binding peptidase activator activity enzyme binding protein kinase binding syntaxin binding protein binding, bridging protein complex scaffold identical protein binding ion channel binding protein heterodimerization activity Rac GTPase binding nitric-oxide synthase binding ATPase binding inward rectifier potassium channel inhibitor activity MDM2/MDM4 family protein binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter MAPK cascade inactivation of MAPK activity angiogenesis vasculogenesis response to hypoxia negative regulation of endothelial cell proliferation positive regulation of endothelial cell proliferation negative regulation of cytokine-mediated signaling pathway response to ischemia regulation of the force of heart contraction by chemical signal triglyceride metabolic process calcium ion transport cellular calcium ion homeostasis regulation of smooth muscle contraction skeletal muscle tissue development response to nutrient lactation protein localization response to bacterium response to gamma radiation positive regulation of calcium ion transport into cytosol posttranscriptional regulation of gene expression positive regulation of gene expression negative regulation of muscle cell apoptotic process positive regulation of peptidase activity vesicle organization receptor-mediated endocytosis of virus by host cell regulation of fatty acid metabolic process lipid storage regulation of blood coagulation cholesterol transport negative regulation of transforming growth factor beta receptor signaling pathway negative regulation of epithelial cell differentiation mammary gland development organ regeneration positive regulation of microtubule polymerization T cell costimulation negative regulation of protein ubiquitination positive regulation of protein ubiquitination receptor internalization negative regulation of protein binding positive regulation of protein binding maintenance of protein location in cell response to progesterone negative regulation of peptidyl-serine phosphorylation positive regulation of peptidyl-serine phosphorylation cholesterol efflux nitric oxide homeostasis positive regulation of toll-like receptor 3 signaling pathway wound healing vasoconstriction response to drug negative regulation of tyrosine phosphorylation of Stat5 protein cholesterol homeostasis positive regulation of catalytic activity negative regulation of MAPK cascade response to estrogen protein localization to plasma membrane raft negative regulation of nitric oxide biosynthetic process negative regulation of neuron differentiation positive regulation of endocytosis positive regulation of vasoconstriction negative regulation of JAK-STAT cascade microtubule polymerization negative regulation of pinocytosis negative regulation of smooth muscle cell proliferation leukocyte migration regulation of nitric-oxide synthase activity negative regulation of nitric-oxide synthase activity protein homooligomerization response to glucocorticoid regulation of cytosolic calcium ion concentration response to calcium ion membrane depolarization regulation of peptidase activity calcium ion homeostasis mammary gland involution positive regulation of cell adhesion molecule production negative regulation of necroptotic process negative regulation of protein tyrosine kinase activity negative regulation of ERK1 and ERK2 cascade caveola assembly cellular response to mechanical stimulus cellular response to exogenous dsRNA cellular response to peptide hormone stimulus cellular response to hyperoxia cellular response to transforming growth factor beta stimulus caveolin-mediated endocytosis regulation of heart rate by cardiac conduction angiotensin-activated signaling pathway involved in heart process negative regulation of canonical Wnt signaling pathway positive regulation of canonical Wnt signaling pathway cellular senescence apoptotic signaling pathway regulation of membrane repolarization during action potential regulation of cardiac muscle cell action potential involved in regulation of contraction regulation of ventricular cardiac muscle cell action potential regulation of ruffle assembly negative regulation of peptidyl-tyrosine autophosphorylation negative regulation of potassium ion transmembrane transport regulation of cell communication by electrical coupling involved in cardiac conduction regulation of inward rectifier potassium channel activity positive regulation of ER-associated ubiquitin-dependent protein catabolic process protein localization to basolateral plasma membrane positive regulation of gap junction assembly beta-catenin destruction complex disassembly receptor internalization involved in canonical Wnt signaling pathway regulation of entry of bacterium into host cell negative regulation of anoikis positive regulation of extrinsic apoptotic signaling pathway positive regulation of intrinsic apoptotic signaling pathway negative regulation of cation channel activity
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-k2m5x:80/100.66.128.182:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0