Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CDC42 Antibodies

          Invitrogen

          Cdc42 Recombinant Rabbit Monoclonal Antibody (23GB1025)

          Advanced Verification
          View all (16) CDC42 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Cdc42 Recombinant Rabbit Monoclonal Antibody (23GB1025)

          • Antibody Testing Data (3)
          • Advanced Verification (1)
          Cdc42 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Cdc42 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Cdc42 Antibody (MA5-53948) in ICC/IF

          Immunocytochemistry analysis of CDC42 in HepG2 cells. Nuclei were stained blue with DAPI; CDC42 recombinant monoclonal antibody (Product # MA5-53948) with a dilution of 1:1,000 was stained magenta with Alexa Fluor 647. Images were taken using Leica stellaris 5. Protein abundance based on laser Intensity and smart gain: Medium. Scale bar: 20 μm. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Cdc42 Antibody in Immunocytochemistry (ICC/IF)
          Cdc42 Antibody in Western Blot (WB)
          Cdc42 Antibody in Flow Cytometry (Flow)
          Cdc42 Antibody

          Product Details

          MA5-53948

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:5,000
          -

          Immunocytochemistry (ICC/IF)

          1:1,000
          -

          Flow Cytometry (Flow)

          1:2,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293F

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          23GB1025

          Immunogen

          Recombinant protein of human CDC42 (60-191AA).
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3248419

          Target Information

          Rac1 is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for the Rac1 gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cell division control protein 42 homolog; cell division cycle 42 (GTP binding protein); cell division cycle 42 homolog; dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein, 25kD)); dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein, 25kD)); G25K GTP-binding protein; growth-regulating protein; GTP binding protein, 25kDa; GTP-binding protein; RP1-224A6.5; small GTP binding protein CDC42

          View more View less

          Gene Aliases: AI747189; AU018915; CDC42; CDC42Hs; G25K; TKS

          View more View less

          UniProt ID: (Human) P60953, (Rat) Q8CFN2, (Mouse) P60766

          View more View less

          Entrez Gene ID: (Human) 998, (Rat) 64465, (Mouse) 12540

          View more View less

          Function(s)
          GTPase activity protein binding GTP binding protein kinase binding GTP-dependent protein binding mitogen-activated protein kinase kinase kinase binding thioesterase binding apolipoprotein A-I receptor binding identical protein binding Rho GDP-dissociation inhibitor binding ubiquitin protein ligase activity nucleotide binding
          Process(es)
          positive regulation of protein phosphorylation sprouting angiogenesis cardiac conduction system development keratinocyte development endocytosis actin filament organization Golgi organization regulation of mitotic nuclear division nuclear migration small GTPase mediated signal transduction metabolic process response to glucose positive regulation of gene expression negative regulation of gene expression single organismal cell-cell adhesion regulation of exocytosis substantia nigra development establishment of cell polarity actin cytoskeleton organization hair follicle morphogenesis positive regulation of pseudopodium assembly negative regulation of protein complex assembly keratinization regulation of protein stability positive regulation of cytokinesis positive regulation of peptidyl-serine phosphorylation adherens junction organization cellular protein localization establishment or maintenance of apical/basal cell polarity multicellular organism growth dendritic cell migration regulation of protein catabolic process positive regulation of catalytic activity positive regulation of MAPK cascade regulation of protein heterodimerization activity positive regulation of neuron apoptotic process positive regulation of phosphatidylinositol 3-kinase activity positive regulation of DNA replication regulation of protein kinase activity positive regulation of JNK cascade filopodium assembly neuron fate determination epidermis morphogenesis actin filament bundle assembly regulation of protein metabolic process regulation of filopodium assembly nucleus localization establishment of Golgi localization positive regulation of synapse structural plasticity regulation of attachment of spindle microtubules to kinetochore heart contraction canonical Wnt signaling pathway submandibular salivary gland formation epithelial-mesenchymal cell signaling hair follicle placode formation positive regulation of hair follicle cell proliferation organelle transport along microtubule actin filament branching epithelial cell-cell adhesion positive regulation of intracellular protein transport positive regulation of substrate adhesion-dependent cell spreading establishment or maintenance of cell polarity blood coagulation protein ubiquitination macrophage differentiation positive regulation of cell growth T cell costimulation Fc-gamma receptor signaling pathway involved in phagocytosis negative regulation of epidermal growth factor receptor signaling pathway vascular endothelial growth factor receptor signaling pathway ephrin receptor signaling pathway regulation of small GTPase mediated signal transduction positive regulation of muscle cell differentiation Wnt signaling pathway, planar cell polarity pathway positive regulation of epithelial cell proliferation involved in lung morphogenesis Rho protein signal transduction nervous system development endosomal transport cell differentiation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0