Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Chemerin Antibodies

          Proteintech

          Chemerin Polyclonal Antibody

          5 References
          View all (27) Chemerin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Chemerin Polyclonal Antibody

          • Antibody Testing Data (3)
          Chemerin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Chemerin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Chemerin Antibody (10216-1-AP) in IHC (P)

          Immunohistochemistry of paraffin-embedded human placenta tissue slide using 10216-1-AP (Chemerin antibody) at dilution of 1:200 (under 40x lens) heat mediated antigen retrieved with Tris-EDTA buffer (pH 9). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Chemerin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Chemerin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Chemerin Antibody in Western Blot (WB)

          Product Details

          10216-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200-1:1,000
          View 3 publications 3 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Chemerin Fusion Protein Ag0261 (1-163 aa encoded by BC000069)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.33 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MRRLLIPLA LWLGAVGVGV AELTEAQRRG LQVALEEFHK HPPVQWAFQE TSVESAVDTP FPAGIFVRLE FKLQQTSCRK RDWKKPECKV RPNGRKRKCL ACIKLGSEDK VLGRLVHCPI ETQVLREAEE HQETQCLRVQ RAGEDPHSFY FPGQFAFSKA LPRS (1-163 aa encoded by BC000069)

          Target Information

          Retinoids act through ligand-dependent transcription factors, retinoid X receptor (RXRs) and retinoic acid receptors (RARs). Tazarotene-induced gene (TIG) proteins, also designated retinoic acid receptor responder proteins or RAR-responsive proteins, can be membrane bound or secreted. TIGs act as tumor suppressor genes in human cancers and are highly expressed in skin, hair follicles and endothelial cells as well as in pancreas, spleen and intestine. TIGs are activated by tazarotene and have been implicated as growth regulators that mediate the growth suppressive effects of retinoids. TIG2 is a secreted protein that is mainly expressed in epidermis, hair follicles and endothelial cells. TIG2 is inhibited in psoriatic lesions and is activated by tazarotene in skin rafts and in epidermis of psoriatic lesions.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Chemerin; RAR-responsive protein TIG2; retinoic acid receptor responder (tazarotene induced) 2; Retinoic acid receptor responder protein 2; Tazarotene-induced gene 2 protein; TIG-2; TIG2HP10433

          View more View less

          Gene Aliases: 0610007L05Rik; AI303516; HP10433; RARRES2; TIG2

          View more View less

          UniProt ID: (Human) Q99969, (Mouse) Q9DD06

          View more View less

          Entrez Gene ID: (Human) 5919, (Mouse) 71660

          View more View less

          Function(s)
          receptor binding protein binding
          Process(es)
          retinoid metabolic process positive regulation of protein phosphorylation chemotaxis defense response inflammatory response positive regulation of macrophage chemotaxis cell differentiation positive regulation of fat cell differentiation embryonic digestive tract development brown fat cell differentiation positive regulation of chemotaxis regulation of lipid catabolic process positive regulation of glucose import in response to insulin stimulus in utero embryonic development platelet degranulation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0