Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • hCG Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.

          Invitrogen

          Chorionic Gonadotropin Monoclonal Antibody (FB12), eBioscience™

          1 Published Figure
          1 Reference
          View all (14) hCG antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Datasheet
          Protocols

          Cite Chorionic Gonadotropin Monoclonal Antibody (FB12), eBioscience™

          • Antibody Testing Data (1)
          • Published Figures (1)
          Chorionic Gonadotropin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Chorionic Gonadotropin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Chorionic Gonadotropin Antibody (14-6508-95) in ICC/IF

          Immunocytochemistry on fixed BeWo cells using 5 µg/mL Mouse IgG1 K Isotype Control (left) or 5 µg/mL Anti-Human Chorionic Gonadotropin Purified (right) followed by Anti-Mouse TRITC.Nuclei are counterstained with DAPI. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Chorionic Gonadotropin Antibody in Immunocytochemistry (ICC/IF)
          Chorionic Gonadotropin Antibody in Immunocytochemistry (ICC/IF)

          Product Details

          14-6508-95

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          5 µg/mL
          -

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-Dependent
          -

          Immunocytochemistry (ICC/IF)

          5 µg/mL
          View 1 publication 1 publication

          ELISA (ELISA)

          Assay-Dependent
          -
          Product Specifications

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          FB12

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Storage conditions

          4°C

          Shipping conditions

          Wet ice

          RRID

          AB_2865208

          Product Specific Information

          Description: This FB12 monoclonal antibody reacts with human Chorionic Gonadotropin (hCG). This secreted glycoprotein is composed of an alpha subunit, which is common to other peptide hormones such as luteinizing hormone (LH), follicle stimulating hormone (FSH) and thyroid stimulating hormone (TSH), and a beta subunit which determines the specificity of the hormone. FB12 recognizes both the intact hCG (composed of the alpha and beta subunits) as well as the free beta subunit, highly expressed in both choriocarcinoma and the serum of pregnant women. hCG is normally expressed by trophoblastoid cells (syncytiotrophoblasts and extravillous cytotrophoblasts) that are responsible for maintenance of the corpus luteum during early gestation. hCG plays an important role in the regulation of angiogenesis through activation of the PI3K/mTOR pathway, resulting in upregulation of vascular endothelial growth factor (VEGF). Ectoptic expression of hCG has also been reported in multiple types of cancer (bladder, lung, breast, pancreas, renal).

          Applications Reported: This FB12 antibody has been reported for use in western blotting, immunohistochemical staining of frozen tissue sections (IHC-F), ELISA, and immunocytochemistry (ICC).

          Applications Tested: This FB12 antibody has been tested by western blot and immunocytochemistry on BeWo cells. This can be used at less than or equal to 5 µg/mL. it is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Human chorionic gonadotropin (hCG) is a placental hormone that stimulates secretion of the pregnancy-sustaining steroid progesterone. hCG is a member of a family of glycoprotein hormones including pituitary hormones lutropin (luteinizing hormone;LH), follitropin (follicle-stimulating hormone; FSH) and thyrotropin (thyroid-stimulating hormone; TSH). Members of this family are all composed of disulfide-rich heterodimers with two dissimilar subunits namely alpha- and beta-subunits. The alpha subunit is common to all the members of the family within a given species. The beta subunits are distinct in each of the horomones and confer the receptor and biological specificity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0