Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The antibody is specific to COL16A1.
This gene encodes the alpha chain of type XVI collagen, a member of the FACIT collagen family (fibril-associated collagens with interrupted helices). Members of this collagen family are found in association with fibril-forming collagens such as type I and II, and serve to maintain the integrity of the extracellular matrix. High levels of type XVI collagen have been found in fibroblasts and keratinocytes, and in smooth muscle and amnion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [a]1 (XVI) collagen; alpha 1 type XVI collagen; col16a1; Collagen alpha-1(XVI) chain; collagen XVI, alpha-1 polypeptide; collagen, type XVI, alpha 1; procollagen type XVI alpha 1; procollagen, type XVI, alpha 1
Gene Aliases: 2700007F12Rik; 447AA; A530052M23Rik; AI838662; CA1F; COL16A1; FP1572
UniProt ID: (Human) Q07092, (Mouse) Q8BLX7
Entrez Gene ID: (Human) 1307, (Rat) 366474, (Mouse) 107581
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support