Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Contactins mediate cell surface interactions during nervous system development. Participates in oligodendrocytes generation by acting as a ligand of NOTCH1. Its association with NOTCH1 promotes NOTCH1 activation through the released notch intracellular domain (NICD) and subsequent translocation to the nucleus. Involved in motor coordination. Tissue specificity: Expressed in nervous system. Highly expressed in cerebellum. Expressed at intermediate level in thalamus, subthalamic nucleus. Weakly expressed in corpus callosum, caudate nucleus and spinal cord. Similarity: Belongs to the immunoglobulin superfamily. Contactin family. Contains 4 fibronectin type-III domains. Contains 6 Ig-like C2-type (immunoglobulin-like) domains.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: axonal-associated cell adhesion molecule; BIG-1; BIG-2; brain-derived immunoglobulin superfamily molecule; Brain-derived immunoglobulin superfamily protein 1; Brain-derived immunoglobulin superfamily protein 2; contactin 3 (plasmacytoma associated); Contactin-3; Contactin-4; Contactin-5; Contactin-6; hNB-2; hNB-3; mNB-3; neural adhesion molecule; neural cell adhesion protein BIG-2; Neural recognition molecule NB-2; Neural recognition molecule NB-3; Plasmacytoma-associated neuronal glycoprotein; Plasmacytoma-associated neuronal glycoprotein (neural cell adhesion molecule BIG-1)
Gene Aliases: 6720426O10Rik; 9630050B05; A830025P08Rik; AXCAM; BIG-1; BIG-2; CNTN3; CNTN4; CNTN5; CNTN6; Gm507; HNB-2s; KIAA1496; Kiaa3024; NB-2; NB-3; NB3; PANG; PCS
UniProt ID: (Human) Q9P232, (Human) Q8IWV2, (Human) O94779, (Human) Q9UQ52, (Mouse) Q07409, (Rat) Q62682, (Rat) Q62845, (Mouse) Q69Z26, (Mouse) P68500, (Rat) P97527, (Mouse) Q9JMB8, (Rat) P97528
Entrez Gene ID: (Human) 5067, (Human) 152330, (Human) 53942, (Human) 27255, (Mouse) 18488, (Rat) 54279, (Rat) 116658, (Mouse) 269784, (Mouse) 244682, (Rat) 114589, (Mouse) 53870, (Rat) 27256
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support