Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: EPTKPPASIF ARSLSATDIE VFWASPLEKN RGRIQGYEVK YWRHEDKEEN ARKIRTVGNQ TSTKITNLKG SVLYHLAVKA YNSAGTGPSS ATVNVTTRK
The protein encoded by this gene is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: axonal-associated cell adhesion molecule; BIG-2; Brain-derived immunoglobulin superfamily protein 2; CNTN4A; Contactin-4; MGC33615; neural cell adhesion protein BIG-2; SCA16
Gene Aliases: AXCAM; BIG-2; CNTN4
UniProt ID: (Human) Q8IWV2
Entrez Gene ID: (Human) 152330
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support