Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunohistochemistry (PFA fixed): incubate antibody for 30 min RT. Staining of formalin-fixed tissues requires heating tissue sections in 10mM Tris with 1mM EDTA, pH 9.0, for 45 min at 95 degrees C followed by cooling at RT for 20 minutes.
Cytochrome C is an electron transporting protein that resides within the intermembrane space of the mitochondria, where it plays a critical role in oxidative phosphorylation and the production of cellular ATP. Release of cytochrome C from the mitochondria to the cytosol is triggered by apoptotic stimuli. In the cytoplasm, the cytochrome C binds apoptotic protease activating factor which activates the apoptotic initiator procaspase 9. Cytosolic cytochrome C functions in the activation of caspase 3, an ICE family molecule that is a key effector of apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CG17903-PA; CG17903-PB; CRF; Cyt-c-p-PA; Cyt-c-p-PB; Cytochrome c; cytochrome c DC4; cytochrome c proximal; Cytochrome C, expressed in somatic tissues; Cytochrome c, somatic; Cytochrome c-2; cytochrome c-like protein; Cytochrome c-proximal; cytochrome-c-p; Cytochrome-c-proximal; hc1
Gene Aliases: CG17903; CYC; CYCS; CYCSA; Cyct-c-p; Cyt C; cyt c-p; Cyt-c-p; Cyt-c2; cyt.c; CytC; CytC-2; Cytc-p; CYTC2; cytochrome c; DC4; dCyt.C-p; DMc01; Dmel\CG17903; Dmel_CG17903; HCS; pDMc01; THC4
UniProt ID: (Dog) P00011, (Human) P99999, (Mouse) P62897, (Rat) P62898, (Fruit fly) P84029
Entrez Gene ID: (Dog) 475258, (Human) 54205, (Horse) 100053958, (Mouse) 13063, (Rat) 25309, (Fruit fly) 34996
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support