Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Cytochromes P450 are a group of heme-thiolate monooxygenases. In liver microsomes, this enzyme is involved in an NADPH-dependent electron transport pathway. It oxidizes a variety of structurally unrelated compounds, including steroids, fatty acids, and xenobiotics.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CYPIIIA1; cytochrome P-450PCN (PNCN inducible); cytochrome P450 3A1; cytochrome P450, family 3, subfamily a, polypeptide 1; cytochrome P450, subfamily 3A, polypeptide 3; Cytochrome P450, subfamily IIIA, polypeptide 3; cytochrome P450-PCN1; cytochrome P450/6 beta B; P450 IIIA; P450-PCN1
Gene Aliases: cDEX; CYP; Cyp3a1; CYP3A23; Cyp3a3; RL33
Entrez Gene ID: (Rat) 25642
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support