Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Progen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Ks18.04 represents an excellent marker to discriminate simple epithelia from those of different origin. Tumors specifically detected: all adenocarcinoma; mammary carcinoma, urinary bladder carcinoma, undifferentiated carcinoma, cervix carcinoma, hepatocellular carcinoma. Polypeptide reacting: MW 45 000 polypeptide (human keratin K18; formerly also designated cytokeratin 18) of all simple type epithelia and basal cells of many squamous, nonepidermal epithelia.Tested cultured cell lines: MCF-7.
Cytokeratin 18 (Keratin 18, KRT18, type I cytoskeletal 18) belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18. Further, cytokeratin 18 is found primarily in non-squamous epithelia and is present in a majority of adenocarcinomas and ductal carcinomas, but not in squamous cell carcinomas. Hepatocellular carcinomas express only cytokeratins 8 and 18. Mutations in the cytokeratin 18 gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for the cytokeratin 18 gene. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell proliferation-inducing gene 46 protein; CK 18; CK-18; cytokeratin 18; Cytokeratin endo B; cytokeratin type -1; DAPK-1; Cytokeratin-18; Cytokeratin18; Dorsal aorta proneprin kinesin-1; DreK18; Endo B; K 18; K18; keratin 18; keratin 18, type I; keratin complex 1, acidic, gene 18; Keratin D; Keratin, type I cytoskeletal 18; Keratin-18; Keratin18; KRT 18; krt18 {ECO:0000250|UniProtKB:P05783}; type I cytoskeletal 18
Gene Aliases: BOS_5019; cb83; CK-18; CK18; CYK18; dapk1; DreK18; I79_023185; K18; Kerd; Krt1-18; KRT18; PIG46; sb:cb83; wu:fa13f02; wu:fb36b04
UniProt ID: (Human) P05783, (Rat) Q5BJY9, (Mouse) P05784, (Zebrafish) Q7ZTS4
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support