Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total KRT31/33A/33B.
The protein encoded by this gene is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hair keratin; Hair keratin, type I Ha1; Hair keratin, type I Ha3; Hair keratin, type I Ha3-I; Hair keratin, type I Ha3-II; hard keratin, type I, 1; hard keratin, type I, 3II; hard keratin, type I,3I; HKA-1; K31; K33A; K33B; keratin 31, type I; keratin 33A, type I; keratin 33B, type I; keratin complex 1, acidic, gene 1; keratin complex 1, acidic, gene 3; keratin complex 1, gene 1; keratin complex 1, gene 3; keratin, hair, acidic, 3A; keratin, hair, acidic, 3B; keratin, hair, acidic,1; Keratin, type I cuticular Ha1; Keratin, type I cuticular Ha3; Keratin, type I cuticular Ha3-I; Keratin, type I cuticular Ha3-II; Keratin-31; Keratin-33A; Keratin-33B; krt33a; ortholog of keratin, hair, acidic, 3A KRTHA3A; ortholog of keratin, hair, acidic, 3B KRTHA3B; type I hair keratin 3B; type I hair keratin KA25; type I hair keratin KA27; type I hair keratin KA28
Gene Aliases: 2310015J09Rik; Ha-1; Ha-3I; Ha-3II; HA1; Ha3; HA3I; HA3II; Ha4; HHA1; HHA3-I; HHA3-II; HKA1; Hka3; HKA3A; HKA3B; K33A; K33B; Ka25; Ka27; Ka28; Kha1; Krt1-1; Krt1-3; KRT31; KRT33A; KRT33B; KRTHA1; KRTHA3A; KRTHA3B; mHa3; MKHA-1; RGD1306249
UniProt ID: (Human) Q15323, (Human) O76009, (Human) Q14525, (Mouse) Q61765, (Mouse) Q8K0Y2, (Mouse) Q61897
Entrez Gene ID: (Human) 3881, (Human) 3883, (Human) 3884, (Rat) 450228, (Mouse) 16660, (Rat) 303527, (Mouse) 71888, (Rat) 450227, (Mouse) 16671
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support