Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody reacts with cytokeratin peptides 4, 5, 6, 8, 10, 13, 18. Cytokeratins are members of intermediate filaments subfamily intracellular proteins represented in epithelial tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 58 kDa cytokeratin; Cell proliferation-inducing gene 46 protein; CK-10; CK-13; CK-18; CK-4; CK-5; CK-8; cytokeratin 10; cytokeratin 13; cytokeratin 18; cytokeratin 4; Cytokeratin-10; Cytokeratin-13; Cytokeratin-18; Cytokeratin-4; Cytokeratin-5; Cytokeratin-8; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; K10; K13; K18; K4; K5; K8; keratin 10, type I; keratin 13, type I; keratin 18, type I; keratin 4, type II; keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin 8, type II; Keratin, type I cytoskeletal 10; Keratin, type I cytoskeletal 13; Keratin, type I cytoskeletal 18; Keratin, type II cytoskeletal 4; Keratin, type II cytoskeletal 5; Keratin, type II cytoskeletal 8; Keratin-10; Keratin-13; Keratin-18; Keratin-4; Keratin-5; Keratin-8; Type-II keratin Kb4; Type-II keratin Kb5; Type-II keratin Kb8
Gene Aliases: BCIE; BIE; CARD2; CK-18; CK-4; CK-8; CK10; CK13; CK4; CK5; CK8; CYK18; CYK4; CYK8; DDD; DDD1; EBS2; EHK; K10; K13; K18; K2C8; K4; K5; K8; KO; KPP; KRT10; KRT13; KRT18; KRT4; KRT5; KRT5A; KRT8; PIG46; WSN1; WSN2
UniProt ID: (Human) P13645, (Human) P13646, (Human) P05783, (Human) P19013, (Human) P13647, (Human) P05787
Entrez Gene ID: (Human) 3858, (Human) 3860, (Human) 3875, (Human) 3851, (Human) 3852, (Human) 3856
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support