Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MUbio
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
RCK102 is a Cytokeratin antibody reacting with Cytokeratin 5 and Cytokeratin 8. This monoclonal antibody recognizes virtually all epithelial tissues and carcinomas.
The antibody may be stored at 4°C. For prolonged storage prepare appropriate aliquots and store at or below -20°C. Prior to use, an aliquot is thawed slowly in the dark at ambient temperature, spun down again and used to prepare working dilutions by adding sterile phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. Working dilutions should be stored at 4°C, not refrozen, and preferably used the same day. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance or the concentration of the product.
Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between ~40 kDa and ~68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 58 kDa cytokeratin; CK-5; CK-8; cytokeratin 5; cytokeratin 8; Cytokeratin endo A; Cytokeratin-5; Cytokeratin-8; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; K5; K8; keratin 5; keratin 5 (epidermolysis bullosa simplex Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin 5-like; keratin 8; keratin 8, type II; keratin complex 2, basic, gene 5; keratin complex 2, basic, gene 8; Keratin, type II cytoskeletal 5; Keratin, type II cytoskeletal 8; keratin, type II cytoskeletal 8-like protein; Keratin-5; Keratin-8; krt5; tuftelin interacting protein 8; Type-II keratin Kb5; Type-II keratin Kb8
Gene Aliases: 3300001P10Rik; AA960620; AL022697; AU019895; AW146334; CARD2; CK-8; CK5; CK8; CYK8; CYKER; DDD; DDD1; EBS2; EndoA; K2C8; K5; K8; KO; Krt-2.8; Krt2-5; Krt2-8; KRT5; KRT5A; KRT8; Sak57; Tfip8
UniProt ID: (Human) P13647, (Human) P05787, (Rat) Q6P6Q2, (Mouse) Q922U2, (Mouse) P11679, (Rat) Q10758
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support