Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Cytokeratin Pan Type II Antibodies

          NeoBiotechnologies

          Cytokeratin, Basic (Type II or HMW) Monoclonal Antibody (SPM591)

          View all (36) Cytokeratin Pan Type II antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Cytokeratin, Basic (Type II or HMW) Monoclonal Antibody (SPM591)

          • Antibody Testing Data (1)
          Cytokeratin, Basic (Type II or HMW) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Cytokeratin, Basic (Type II or HMW) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Cytokeratin, Basic (Type II or HMW) Antibody (MSM3X-446-P0) in IHC (P)

          Formalin-fixed, paraffin-embedded human Prostate Carcinoma stained with Cytokeratin, HMW Monoclonal Antibody (SPM591). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Cytokeratin, Basic (Type II or HMW) Antibody in Immunohistochemistry (Paraffin) (IHC (P))

          Product Details

          MSM3X-446-P0

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-2 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-dependent
          -

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          1-2 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          1-2 µg/mL
          -

          Flow Cytometry (Flow)

          1-2 µg/10^6 cells
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          SPM591

          Immunogen

          Solubilized keratin extract from human stratum corneum
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          200 µg/mL

          Purification

          Protein A/G

          Storage buffer

          PBS, pH 7.4, with 0.05% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          4°C, do not freeze

          Shipping conditions

          Wet ice

          Target Information

          Cytokeratin pan is part of a subfamily of intermediate filament proteins that are characterized by remarkable biochemical diversity, and represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight between 40 kDa- 68 kDa, and an isoelectric pH between 4.9-7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. The composition of cytokeratin pairs vary with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 57kd keratin; 57kDa keratin; 58 kDa cytokeratin; 60-kDa keratin; 65 kDa cytokeratin; 67 kDa cytokeratin; CK 2e; CK-1; CK-2e; CK-2P; CK-3; CK-4; CK-5; CK-6A; CK-6D; CK-7; CK-72; CK-8; cytokeratin 1; cytokeratin 2; cytokeratin 3; cytokeratin 4; cytokeratin 5; cytokeratin 6A; cytokeratin 6C; cytokeratin 6D; Cytokeratin 7; cytokeratin 8; Cytokeratin endo A; Cytokeratin-1; Cytokeratin-2e; Cytokeratin-2P; Cytokeratin-3; Cytokeratin-4; Cytokeratin-5; Cytokeratin-6A; cytokeratin-6C; Cytokeratin-6D; cytokeratin-6E; Cytokeratin-7; Cytokeratin-72; Cytokeratin-8; Cytoskeletal 57 kDa keratin; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; epidermolytic hyperkeratosis 1; Epithelial keratin-2e; Hair alpha protein; K1; K2e; K2P; K3; K4; K5; K6A; K7; K72; K76; K8; kamp-keratin derived antimicrobial peptide; KDAMP; keratin 1, type II; keratin 2 epidermis; keratin 2, type II; keratin 2p; keratin 3, type II; keratin 4, type II; keratin 5 (epidermolysis bullosa simplex Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin 6, inner root sheath, 2; keratin 6A, , type II; keratin 6A, type II; keratin 7, type II; keratin 72, pseudogene; keratin 72, type II; keratin 76, type II; keratin 8, type II; keratin 86; keratin complex 2, basic, gene 1; keratin complex 2, basic, gene 17; keratin complex 2, basic, gene 35; keratin complex 2, basic, gene 4; keratin complex 2, basic, gene 5; keratin complex 2, basic, gene 6a; keratin complex 2, basic, gene 6c; keratin complex 2, basic, gene 7; keratin complex 2, basic, gene 8; keratin complex 2, gene 17; keratin complex 2, gene 6a; keratin K6h; keratin protein K6irs; keratin, 55K type II cytoskeletal; keratin, epidermal type II, K6A; keratin, simple epithelial type I, K7; Keratin, type II cytoskeletal 1; Keratin, type II cytoskeletal 2 epidermal; Keratin, type II cytoskeletal 2 oral; Keratin, type II cytoskeletal 3; Keratin, type II cytoskeletal 4; Keratin, type II cytoskeletal 5; Keratin, type II cytoskeletal 6A; Keratin, type II cytoskeletal 7; Keratin, type II cytoskeletal 72; Keratin, type II cytoskeletal 8; Keratin-1; Keratin-2 epidermis; Keratin-2e; Keratin-3; Keratin-4; Keratin-5; Keratin-6-alpha; Keratin-6A; keratin-6C; Keratin-7; Keratin-72; Keratin-76; Keratin-8; krt1 {ECO:0000250|UniProtKB:P04264}; krt2; krt2 {ECO:0000250|UniProtKB:P35908}; krt4 {ECO:0000250|UniProtKB:P19013}; krt5; krt7; krt7 {ECO:0000250|UniProtKB:P08729}; krt76; MGC116967; MGC116968; mK6-alpha; pan keratin; Sarcolectin; tuftelin interacting protein 8; Type II inner root sheath-specific keratin-K6irs2; type II keratin Kb1; type II keratin Kb2; type II keratin Kb35; type II keratin Kb4; type II keratin Kb7; type II keratin Kb9; type II mesothelial keratin K7; Type-II keratin Kb1; type-II keratin Kb12; Type-II keratin Kb2; Type-II keratin Kb3; Type-II keratin Kb35; Type-II keratin Kb4; Type-II keratin Kb5; Type-II keratin Kb6; Type-II keratin Kb7; Type-II keratin Kb8; Type-II keratin Kb9; type-II keratin-35

          View more View less

          Gene Aliases: 2310001L23Rik; 2310016L08Rik; 3300001P10Rik; AA960620; AI507495; AL022697; AU019895; AW108092; AW146334; BB005427; CARD2; CK-2e; CK-4; CK-6C; CK-6E; CK-8; CK1; CK3; CK4; CK5; CK6A; CK6C; CK6D; CK7; CK8; CYK4; CYK8; CYKER; D15Wsu77e; DDD; DDD1; EBS2; EHK; EHK1; EndoA; EPPK; HUMCYT2A; K1; K2C7; K2C8; K2e; K3; K4; K5; K6A; K6C; K6D; K6irs; K6IRS2; K7; K8; Kb1; Kb2; KB35; Kb4; Kb7; Kb9; Ker2; KO; Krt-2.1; Krt-2.4; Krt-2.8; KRT1; KRT1A; KRT2; Krt2-1; Krt2-17; Krt2-2; Krt2-35; Krt2-4; Krt2-5; Krt2-6; Krt2-6a; Krt2-6c; Krt2-7; Krt2-8; KRT2A; KRT2B; KRT2E; KRT2P; KRT3; KRT4; KRT5; KRT5A; KRT6; KRT6A; KRT6C; KRT6D; KRT6IRS2; KRT7; KRT72; Krt72-ps; KRT76; KRT8; Krt86; KRTA; KRTE; mK6[a]; MK6a; NEPPK; PC3; Sak57; SCL; Tfip8; WSN1

          View more View less

          UniProt ID: (Human) P04264, (Human) P35908, (Human) P12035, (Human) P19013, (Human) P13647, (Human) P02538, (Human) P08729, (Human) Q14CN4, (Human) Q01546, (Human) P05787, (Mouse) P04104, (Rat) Q6IMF3, (Rat) Q6IG02, (Mouse) Q3TTY5, (Mouse) P07744, (Rat) Q6IG00, (Rat) Q6P6Q2, (Mouse) Q922U2, (Mouse) P50446, (Mouse) Q9DCV7, (Rat) Q6IG12, (Mouse) Q6IME9, (Rat) Q6IG04, (Mouse) Q3UV17, (Rat) Q10758, (Mouse) P11679

          View more View less

          Entrez Gene ID: (Human) 3848, (Human) 3849, (Human) 3850, (Human) 3851, (Human) 3852, (Human) 3853, (Human) 3855, (Human) 140807, (Human) 51350, (Human) 3856, (Mouse) 16678, (Rat) 300250, (Rat) 406228, (Mouse) 16681, (Mouse) 16682, (Rat) 315323, (Rat) 369017, (Mouse) 110308, (Mouse) 16687, (Mouse) 110310, (Rat) 300242, (Mouse) 105866, (Rat) 406227, (Rat) 407757, (Mouse) 77055, (Rat) 25626, (Mouse) 16691

          View more View less

          Function(s)
          receptor activity structural molecule activity protein binding carbohydrate binding structural constituent of cytoskeleton scaffold protein binding molecular_function protein complex binding protein heterodimerization activity
          Process(es)
          epithelial cell differentiation intermediate filament cytoskeleton organization complement activation, lectin pathway retina homeostasis response to oxidative stress fibrinolysis regulation of angiogenesis negative regulation of inflammatory response establishment of skin barrier intermediate filament-based process cytoskeleton organization embryo development negative regulation of epithelial cell proliferation biological_process epidermis development sebaceous gland development hair follicle morphogenesis keratinization keratinocyte activation keratinocyte proliferation intermediate filament organization keratinocyte migration tumor necrosis factor-mediated signaling pathway sarcomere organization response to hydrostatic pressure response to other organism cell differentiation involved in embryonic placenta development extrinsic apoptotic signaling pathway hepatocyte apoptotic process viral process cell morphogenesis involved in differentiation multicellular organism development morphogenesis of an epithelium Wnt signaling pathway wound healing hemidesmosome assembly
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0