Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Cytokeratin 4/5/6/8/10/13/18 Antibodies

          NeoBiotechnologies

          Cytokeratin, Multi Monoclonal Antibody (SPM583)

          View all (5) Cytokeratin 4/5/6/8/10/13/18 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Cytokeratin, Multi Monoclonal Antibody (SPM583)

          • Antibody Testing Data (1)
          Cytokeratin, Multi Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Cytokeratin, Multi Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Cytokeratin, Multi Antibody (MSM1X-393-P1ABX) in IHC (P)

          Formalin-fixed, paraffin-embedded colon (10X) stained with Multi Keratin Monoclonal Antibody (SPM583). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Cytokeratin, Multi Antibody in Immunohistochemistry (Paraffin) (IHC (P))

          Product Details

          MSM1X-393-P1ABX

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-2 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-dependent
          -

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          1-2 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          1-4 µg/mL
          -

          Flow Cytometry (Flow)

          1-2 µg/10^6 cells
          -
          Product Specifications

          Species Reactivity

          Amphibian, Bovine, Guinea pig, Goat, Human, Mouse, Non-human primate, Pig, Rat

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          SPM583

          Immunogen

          Keratin-enriched preparation from cultured human A431
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A/G

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 47 kDa cytokeratin; 56 kDa cytokeratin; 57kd keratin; 57kDa keratin; 58 kDa cytokeratin; Cell proliferation-inducing gene 46 protein; CK-10; CK-13; CK-18; CK-4; CK-5; CK-8; cytokeratin 10; cytokeratin 13; cytokeratin 18; cytokeratin 4; cytokeratin 5; cytokeratin 8; Cytokeratin endo A; Cytokeratin endo B; Cytokeratin VIB; Cytokeratin-10; Cytokeratin-13; Cytokeratin-18; Cytokeratin-4; Cytokeratin-5; Cytokeratin-6B; Cytokeratin-8; Cytokeratin-A; Cytoskeletal 57 kDa keratin; Endo B; epidermal keratin 10; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; epidermolytic hyperkeratosis; keratosis palmaris et plantaris; intermediate filament protein; K10; K13; K18; K4; K5; K8; KERA; keratin 10 (epidermolytic hyperkeratosis); keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris); keratin 10, type I; keratin 13, type I; keratin 18, type I; keratin 4, type II; keratin 5 (epidermolysis bullosa simplex Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin 8, type II; keratin complex 1, acidic, gene 10; keratin complex 1, acidic, gene 13; keratin complex 1, acidic, gene 18; keratin complex 2, basic, gene 4; keratin complex 2, basic, gene 5; keratin complex 2, basic, gene 8; Keratin D; Keratin, type I cytoskeletal 10; Keratin, type I cytoskeletal 13; Keratin, type I cytoskeletal 18; Keratin, type I cytoskeletal 59 kDa; Keratin, type II cytoskeletal 4; Keratin, type II cytoskeletal 5; keratin, type II cytoskeletal 60 kDa, component III; Keratin, type II cytoskeletal 8; Keratin-10; Keratin-13; Keratin-18; Keratin-4; Keratin-5; Keratin-8; krt10 {ECO:0000250|UniProtKB:P13645}; krt13 {ECO:0000250|UniProtKB:P13646}; krt18 {ECO:0000250|UniProtKB:P05783}; krt4 {ECO:0000250|UniProtKB:P19013}; krt5; suprabasal cytokeratin 10; tuftelin interacting protein 8; type I keratin KA10; Type I keratin Ka13; type II keratin Kb4; Type-II keratin Kb4; Type-II keratin Kb5; Type-II keratin Kb8

          View more View less

          Gene Aliases: 3300001P10Rik; AA960620; AL022697; AU019895; AW108092; AW146334; BCIE; BIE; BOS_19066; BOS_5003; BOS_5019; BOS_5040; BOS_5041; CARD2; CK-18; CK-4; CK-8; CK10; CK13; CK18; CK4; CK5; CK8; CYK18; CYK4; CYK8; CYKER; D130054E02Rik; DDD; DDD1; EBS2; EHK; EndoA; K10; K13; K18; K1C1; K2C8; K4; K5; K8; Ka10; Ka13; Kb4; Kerd; KO; KPP; Krt-1.10; Krt-1.13; Krt-2.4; Krt-2.8; Krt1-10; Krt1-13; Krt1-18; KRT10; KRT13; KRT18; Krt2-4; Krt2-5; KRT2-8; KRT4; KRT5; KRT5A; KRT8; PIG46; Sak57; Tfip8; WSN1; WSN2

          View more View less

          UniProt ID: (Bovine) P06394, (Human) P13645, (Human) P13646, (Human) P05783, (Human) P19013, (Bovine) Q5XQN5, (Human) P13647, (Bovine) P05786, (Human) P05787, (Mouse) P02535, (Mouse) P08730, (Rat) Q6IFV4, (Rat) Q5BJY9, (Mouse) P05784, (Rat) Q6IG00, (Mouse) P07744, (Rat) Q6P6Q2, (Mouse) Q922U2, (Mouse) P11679, (Rat) Q10758

          View more View less

          Entrez Gene ID: (Bovine) 281888, (Human) 3858, (Human) 3860, (Human) 3875, (Bovine) 506480, (Human) 3851, (Bovine) 527819, (Bovine) 281268, (Human) 3852, (Bovine) 281269, (Pig) 100152077, (Human) 3856, (Mouse) 16661, (Rat) 450225, (Mouse) 16663, (Rat) 287699, (Rat) 294853, (Mouse) 16668, (Rat) 315323, (Mouse) 16682, (Rat) 369017, (Mouse) 110308, (Mouse) 16691, (Rat) 25626

          View more View less

          Function(s)
          structural molecule activity structural constituent of epidermis protein binding poly(A) RNA binding scaffold protein binding cadherin binding involved in cell-cell adhesion structural constituent of cytoskeleton protein complex binding molecular_function protein heterodimerization activity
          Process(es)
          cytoskeleton organization epithelial cell differentiation negative regulation of epithelial cell proliferation cellular response to calcium ion cell morphogenesis involved in differentiation multicellular organism development tumor necrosis factor-mediated signaling pathway sarcomere organization response to hydrostatic pressure response to other organism cell differentiation involved in embryonic placenta development extrinsic apoptotic signaling pathway hepatocyte apoptotic process cell cycle anatomical structure morphogenesis viral process Golgi to plasma membrane CFTR protein transport negative regulation of apoptotic process intermediate filament cytoskeleton organization cell-cell adhesion response to radiation tongue morphogenesis cellular response to retinoic acid biological_process embryo development keratinocyte differentiation epidermis development hemidesmosome assembly
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0