Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Cytokeratin Pan Type I/II Antibodies

          NeoBiotechnologies

          Cytokeratin, Multi (Epithelial Marker) Monoclonal Antibody (rKRT/457)

          View all (71) Cytokeratin Pan Type I/II antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Cytokeratin, Multi (Epithelial Marker) Monoclonal Antibody (rKRT/457)

          • Antibody Testing Data (2)
          Cytokeratin, Multi (Epithelial Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Cytokeratin, Multi (Epithelial Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Cytokeratin, Multi (Epithelial Marker) Antibody (MSM30-1876-P1ABX) in IHC (P)

          Formalin-fixed, paraffin-embedded human Skin stained with Multi-Cytokeratin Mouse Recombinant Monoclonal Antibody (rKRT/457). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Cytokeratin, Multi (Epithelial Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cytokeratin, Multi (Epithelial Marker) Antibody in SDS-PAGE (SDS-PAGE)

          Product Details

          MSM30-1876-P1ABX

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-dependent
          -

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          1-2 µg/mL
          -
          Product Specifications

          Species Reactivity

          Goat, Human, Mouse, Non-human primate, Pig, Rat, Amphibian, Bovine, Guinea pig

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          rKRT/457

          Immunogen

          Keratin-enriched preparation from cultured human epithelial cells
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A/G

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          Cytokeratin pan is part of a subfamily of intermediate filament proteins that are characterized by remarkable biochemical diversity, and represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight between 40 kDa- 68 kDa, and an isoelectric pH between 4.9-7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. The composition of cytokeratin pairs vary with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 39.1; 40-kDa keratin intermediate filament; 47 kDa cytokeratin; 56 kDa cytokeratin; 57kd keratin; 57kDa keratin; 58 kDa cytokeratin; 60-kDa keratin; 65 kDa cytokeratin; 67 kDa cytokeratin; Cell proliferation-inducing gene 46 protein; CK 2e; CK-1; CK-10; CK-13; CK-14; CK-15; CK-16; CK-17; CK-18; CK-19; CK-1B; CK-2e; CK-2P; CK-3; CK-4; CK-5; CK-6A; CK-6D; CK-7; CK-8; cytokeratin 1; cytokeratin 10; cytokeratin 13; cytokeratin 14; cytokeratin 15; cytokeratin 16; cytokeratin 18; cytokeratin 19; cytokeratin 2; cytokeratin 3; cytokeratin 4; cytokeratin 5; cytokeratin 6A; cytokeratin 6C; cytokeratin 6D; Cytokeratin 7; cytokeratin 8; Cytokeratin endo A; Cytokeratin endo B; Cytokeratin VIB; cytokeratin VII; Cytokeratin-1; Cytokeratin-10; Cytokeratin-13; Cytokeratin-14; Cytokeratin-15; Cytokeratin-16; Cytokeratin-17; Cytokeratin-18; Cytokeratin-19; Cytokeratin-1B; Cytokeratin-2e; Cytokeratin-2P; Cytokeratin-3; Cytokeratin-4; Cytokeratin-5; Cytokeratin-6A; Cytokeratin-6B; cytokeratin-6C; Cytokeratin-6D; cytokeratin-6E; Cytokeratin-7; Cytokeratin-8; Cytokeratin-A; Cytoskeletal 57 kDa keratin; embryonic type II keratin 1; Embryonic type II keratin-1; Endo B; epidermal keratin 10; epidermal keratin VII; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; epidermolysis bullosa simplex, Dowling-Meara, Koebner; epidermolytic hyperkeratosis 1; epidermolytic hyperkeratosis; keratosis palmaris et plantaris; Epithelial keratin-2e; focal non-epidermolytic palmoplantar keratoderma; Hair alpha protein; HMWCK; intermediate filament protein; K1; K10; K13; K14; K15; K16; K17; K18; K19; K1B; K2e; K2P; K3; K4; K5; K6A; K7; K76; K77; K8; kamp-keratin derived antimicrobial peptide; KDAMP; KERA; keratin 1, type II; keratin 10 (epidermolytic hyperkeratosis); keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris); keratin 10, type I; keratin 13, type I; keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner); keratin 14, type I; keratin 15; keratin 15, type I; keratin 16, type I; keratin 17, type I; keratin 18, type I; keratin 19, type I; Keratin 1B; keratin 2 (epidermal ichthyosis bullosa of Siemens); keratin 2 epidermis; keratin 2, type II; keratin 2A; keratin 2p; keratin 3, type II; keratin 4, type II; keratin 5 (epidermolysis bullosa simplex Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin 6 alpha; keratin 6A, , type II; keratin 6A, type II; keratin 7, type II; keratin 76, type II; keratin 77, type II; keratin 8, type II; keratin 86; keratin complex 1, acidic, gene 10; keratin complex 1, acidic, gene 13; keratin complex 1, acidic, gene 14; keratin complex 1, acidic, gene 15; keratin complex 1, acidic, gene 16; keratin complex 1, acidic, gene 17; keratin complex 1, acidic, gene 18; keratin complex 1, acidic, gene 19; keratin complex 2, basic, gene 1; keratin complex 2, basic, gene 17; keratin complex 2, basic, gene 39; keratin complex 2, basic, gene 4; keratin complex 2, basic, gene 5; keratin complex 2, basic, gene 6a; keratin complex 2, basic, gene 6c; keratin complex 2, basic, gene 7; keratin complex 2, basic, gene 8; keratin complex 2, gene 17; keratin complex 2, gene 6a; Keratin D; keratin intermediate filament 16a; keratin intermediate filament 16b; keratin IV; keratin K6h; Keratin type II cytoskeletal 5; keratin, 55K type II cytoskeletal; keratin, epidermal type II, K6A; keratin, simple epithelial type I, K7; Keratin, type I cytoskeletal 10; Keratin, type I cytoskeletal 13; Keratin, type I cytoskeletal 14; Keratin, type I cytoskeletal 15; Keratin, type I cytoskeletal 16; Keratin, type I cytoskeletal 17; Keratin, type I cytoskeletal 18; Keratin, type I cytoskeletal 19; Keratin, type I cytoskeletal 59 kDa; keratin, type I, 40-kd; Keratin, type II cytoskeletal 1; Keratin, type II cytoskeletal 1b; Keratin, type II cytoskeletal 2 epidermal; Keratin, type II cytoskeletal 2 oral; Keratin, type II cytoskeletal 3; Keratin, type II cytoskeletal 4; Keratin, type II cytoskeletal 5; keratin, type II cytoskeletal 60 kDa, component III; Keratin, type II cytoskeletal 6A; Keratin, type II cytoskeletal 7; Keratin, type II cytoskeletal 8; Keratin-1; Keratin-10; Keratin-13; Keratin-14; Keratin-15; keratin-15, basic; keratin-15, beta; Keratin-16; Keratin-17; Keratin-18; Keratin-19; Keratin-2 epidermis; Keratin-2e; Keratin-3; Keratin-4; Keratin-5; Keratin-6-alpha; Keratin-6A; keratin-6C; Keratin-7; Keratin-76; Keratin-77; Keratin-8; krt1 {ECO:0000250|UniProtKB:P04264}; krt10 {ECO:0000250|UniProtKB:P13645}; krt13 {ECO:0000250|UniProtKB:P13646}; krt15 {ECO:0000250|UniProtKB:P19012}; krt17 {ECO:0000250|UniProtKB:Q04695}; krt18 {ECO:0000250|UniProtKB:P05783}; KRT1B; krt2; krt2 {ECO:0000250|UniProtKB:P35908}; krt4 {ECO:0000250|UniProtKB:P19013}; krt5; krt7; krt7 {ECO:0000250|UniProtKB:P08729}; krt76; mK6-alpha; pan Cytokeratin; pan keratin; pankeratin; Sarcolectin; suprabasal cytokeratin 10; tuftelin interacting protein 8; type I cytoskeletal 15; type I keratin KA10; Type I keratin Ka13; Type I keratin Ka14; Type I keratin Ka15; type I keratin KA16; Type I keratin Ka17; Type I keratin Ka19; type II cytoskeletal 1b; type II keratin Kb1; type II keratin Kb2; type II keratin Kb39; type II keratin Kb4; type II keratin Kb7; type II keratin Kb9; type II mesothelial keratin K7; type-II intermediate filament protein; Type-II keratin Kb1; type-II keratin Kb12; Type-II keratin Kb2; Type-II keratin Kb3; Type-II Keratin Kb39; Type-II keratin Kb4; Type-II keratin Kb5; Type-II keratin Kb6; Type-II keratin Kb7; Type-II keratin Kb8; Type-II keratin Kb9

          View more View less

          Gene Aliases: 2310001L23Rik; 2310016L08Rik; 3300001P10Rik; 39.1; 4732484G22Rik; AA960620; AI324768; AI528832; AI626930; AI663979; AL022697; AU019895; AW108092; AW146334; BB005427; BCIE; BIE; BOS_19066; BOS_19067; BOS_19076; BOS_19084; BOS_19102; BOS_5003; BOS_5010; BOS_5013; BOS_5019; BOS_5035; BOS_5040; BOS_5041; CARD2; CK-17; CK-18; CK-2e; CK-4; CK-6C; CK-6E; CK-8; CK1; CK10; CK13; CK14; CK15; CK16; CK18; CK19; CK3; CK4; CK5; CK6A; CK6C; CK6D; CK7; CK8; CYK18; CYK4; CYK8; CYKER; D130054E02Rik; D15Wsu77e; DDD; DDD1; EBS2; EBS3; EBS4; EHK; EHK1; EndoA; EndoC; EPPK; FNEPPK; HUMCYT2A; K1; K10; K13; K14; K15; K16; K17; K18; K19; K1B; K1C1; K1CO; K1CP; K1CS; K1emb; K2C7; K2C8; K2e; K3; K4; K5; K6A; K6C; K6D; K7; K8; Ka10; Ka13; Ka14; Ka15; Ka16; Ka17; Ka19; Kb1; Kb2; Kb39; Kb4; Kb7; Kb9; Ker2; Kerd; KERIA; KERIB; KO; KPP; Krt-1.10; Krt-1.13; Krt-1.14; Krt-1.19; Krt-2.1; Krt-2.4; Krt-2.8; KRT1; Krt1-10; Krt1-13; Krt1-14; Krt1-15; Krt1-16; Krt1-17; Krt1-18; Krt1-19; KRT10; KRT13; KRT14; KRT15; KRT16; KRT16A; KRT17; KRT18; KRT19; KRT1A; KRT1B; KRT2; Krt2-1; Krt2-17; Krt2-2; Krt2-4; Krt2-5; Krt2-6; Krt2-6a; Krt2-6c; Krt2-7; KRT2-8; Krt239; KRT2A; KRT2B; KRT2E; KRT2P; KRT3; KRT4; KRT5; KRT5A; Krt6; KRT6A; KRT6C; KRT6D; KRT7; KRT76; KRT77; KRT8; Krt86; KRTA; KRTB; KRTE; mK1b; mK6[a]; MK6a; NEPPK; NFJ; PC; PC1; PC2; PC3; PCHC1; PIG46; Sak57; SCL; Tfip8; WSN1; WSN2

          View more View less

          UniProt ID: (Human) P04264, (Bovine) P06394, (Human) P13645, (Human) P13646, (Human) P02533, (Human) P19012, (Human) P08779, (Human) Q04695, (Bovine) A1L595, (Human) P05783, (Bovine) P08728, (Human) P08727, (Human) P35908, (Human) P12035, (Human) P19013, (Bovine) Q5XQN5, (Human) P13647, (Human) P02538, (Human) P08729, (Bovine) Q29S21, (Human) Q01546, (Human) Q7Z794, (Bovine) P05786, (Human) P05787, (Rat) Q6IMF3, (Mouse) P04104, (Mouse) P02535, (Mouse) P08730, (Rat) Q6IFV4, (Rat) Q6IFV1, (Mouse) Q61781, (Mouse) Q61414, (Rat) Q6IFV3, (Mouse) Q9Z2K1, (Mouse) Q9QWL7, (Rat) Q6IFU8, (Mouse) P05784, (Rat) Q5BJY9, (Mouse) P19001, (Rat) Q63279, (Mouse) Q3TTY5, (Rat) Q6IG02, (Mouse) P07744, (Rat) Q6IG00, (Rat) Q6P6Q2, (Mouse) Q922U2, (Mouse) P50446, (Mouse) Q9DCV7, (Rat) Q6IG12, (Mouse) Q3UV17, (Rat) Q6IG01, (Mouse) Q6IFZ6, (Rat) Q10758, (Mouse) P11679

          View more View less

          Entrez Gene ID: (Human) 3848, (Bovine) 281888, (Human) 3858, (Human) 3860, (Human) 3861, (Bovine) 404111, (Bovine) 507464, (Human) 3866, (Human) 3868, (Human) 3872, (Bovine) 281889, (Human) 3875, (Bovine) 506480, (Bovine) 514812, (Human) 3880, (Human) 3849, (Pig) 100049665, (Human) 3850, (Bovine) 407188, (Bovine) 527819, (Human) 3851, (Bovine) 281268, (Human) 3852, (Bovine) 614456, (Human) 3853, (Human) 3855, (Bovine) 535697, (Human) 51350, (Bovine) 407189, (Human) 374454, (Bovine) 281269, (Human) 3856, (Pig) 100152077, (Rat) 300250, (Mouse) 16678, (Rat) 450225, (Mouse) 16661, (Mouse) 16663, (Rat) 287699, (Rat) 287701, (Mouse) 16664, (Mouse) 16665, (Rat) 287700, (Rat) 303530, (Mouse) 16666, (Mouse) 16667, (Rat) 287702, (Mouse) 16668, (Rat) 294853, (Mouse) 16669, (Rat) 360626, (Mouse) 16681, (Rat) 406228, (Mouse) 16682, (Rat) 315323, (Rat) 369017, (Mouse) 110308, (Mouse) 16687, (Mouse) 110310, (Rat) 300242, (Rat) 407757, (Mouse) 77055, (Rat) 406226, (Mouse) 406220, (Rat) 25626, (Mouse) 16691

          View more View less

          Function(s)
          receptor activity structural molecule activity protein binding carbohydrate binding structural constituent of epidermis structural constituent of cytoskeleton keratin filament binding scaffold protein binding MHC class II receptor activity MHC class II protein binding poly(A) RNA binding cadherin binding involved in cell-cell adhesion structural constituent of muscle protein complex binding molecular_function protein heterodimerization activity
          Process(es)
          epithelial cell differentiation intermediate filament cytoskeleton organization morphogenesis of an epithelium signal transduction positive regulation of cell growth hair follicle morphogenesis keratinization intermediate filament organization positive regulation of translation positive regulation of hair follicle development cytoskeleton organization response to estrogen sarcomere organization cell differentiation involved in embryonic placenta development cell cycle anatomical structure morphogenesis viral process tumor necrosis factor-mediated signaling pathway Golgi to plasma membrane CFTR protein transport negative regulation of apoptotic process extrinsic apoptotic signaling pathway hepatocyte apoptotic process cell-cell adhesion aging response to zinc ion response to ionizing radiation hair cycle intermediate filament bundle assembly complement activation, lectin pathway retina homeostasis biological_process negative regulation of inflammatory response establishment of skin barrier keratinocyte differentiation cellular response to calcium ion negative regulation of epithelial cell proliferation Notch signaling pathway epidermis development intermediate filament-based process inflammatory response cell proliferation negative regulation of cell migration innate immune response keratinocyte migration sebaceous gland development response to radiation tongue morphogenesis cellular response to retinoic acid response to hydrostatic pressure response to other organism hemidesmosome assembly response to oxidative stress fibrinolysis regulation of angiogenesis mitophagy in response to mitochondrial depolarization embryo development keratinocyte activation keratinocyte proliferation immune system process cell morphogenesis involved in differentiation multicellular organism development regulation of translation Wnt signaling pathway wound healing
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0