Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
The antibody detects a 125 kDa* protein corresponding to the apparent molecular mass of DAAM1 on SDS-PAGE immunoblots of human A431, K562, and mouse C2C12 cells.
DAAM1 is a diaphanous-related formin first studied as a novel Disheveled binding protein and shown to be crucial for the planar cell polarity pathway in Xenopus. DAAM1, like other formins, directs nucleation and elongation of new actin filaments using its conserved formin-homology-2 (FH2) domain. It is a Rho-regulated formin implicated in neuronal development and regulation of gastrulation. It was identified as a binding protein of the Wnt receptor-associated protein Disheveled. It also binds the GTP-bound RhoA and increases its GTP-bound form population in cells. It exists in an autoinhibited state via intramolecular interactions between its amino-terminal GBD and carboxyl-terminal DAD domains. Structurally the protein includes a GTPase-binding domain (GBD), a diaphanous inhibitory domain (DID), and a coiled-coil region followed by the FH1, FH2 and C-terminal diaphanous auto-regulatory domain (DAD). In humans, the DAAM1 gene is mapped to chromosome 14q23.1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Disheveled-associated activator of morphogenesis 1
Gene Aliases: DAAM1; KIAA0666
UniProt ID: (Human) Q9Y4D1, (Mouse) Q8BPM0
Entrez Gene ID: (Human) 23002, (Mouse) 208846, (Rat) 314212
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support