Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • DAB2IP Antibodies

          Invitrogen

          DAB2IP Monoclonal Antibody (1626CT702.263.21)

          View all (10) DAB2IP antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite DAB2IP Monoclonal Antibody (1626CT702.263.21)

          • Antibody Testing Data (1)
          DAB2IP Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DAB2IP Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DAB2IP Antibody (MA5-37700) in WB

          Western Blot analysis of DAB2IP using the DAB2IP Monoclonal Antibody (Product # MA5-37700) at a 1:1000 dilution. Lane 1: Du145 whole cell lysate, Lane 2: HepG2 whole cell lysate, Lane 3: mouse brain lysate, Lane 4: Hela whole cell lysate. Lysates/proteins at 20 µg per lane. Secondary Goat Anti-mouse IgG, (H+L), Peroxidase conjugated at 1:10,000 dilution. Predicted band size : 132 kDa. Blocking/Dilution buffer: 5% NFDM/TBST. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DAB2IP Antibody in Western Blot (WB)

          Product Details

          MA5-37700

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Mouse / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1626CT702.263.21

          Immunogen

          recombinant protein between 782-1038 amino acids from human DAB2IP
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.45 mg/mL

          Purification

          Protein G

          Storage buffer

          PBS, pH 7.4

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2897624

          Target Information

          DAB2IP is a Ras GTPase-activating protein (GAP) that acts as a tumor suppressor. The DAB2IP gene is inactivated by methylation in prostate and breast cancers.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: AIP-1; ASK-interacting protein 1; ASK1-interacting protein 1; DAB2 interaction protein; DAB2-interacting protein; disabled 2 interacting protein long form; Disabled homolog 2-interacting protein; DOC-2/DAB-2 interactive protein; DOC-2/DAB2 interactive protein; nGAP-like protein

          View more View less

          Gene Aliases: 2310011D08Rik; AF9Q34; AI480459; AIP-1; AIP1; DAB2IP; DIP1/2; KIAA1743; mKIAA1743

          View more View less

          UniProt ID: (Human) Q5VWQ8, (Mouse) Q3UHC7

          View more View less

          Entrez Gene ID: (Human) 153090, (Mouse) 69601

          View more View less

          Function(s)
          GTPase activator activity death receptor binding protein binding SH3 domain binding kinase binding protein kinase binding mitogen-activated protein kinase kinase binding mitogen-activated protein kinase kinase kinase binding phosphatidylinositol-3-phosphate binding protein complex binding signaling adaptor activity Toll-like receptor 4 binding phosphatidylinositol 3-kinase regulatory subunit binding identical protein binding protein homodimerization activity vascular endothelial growth factor receptor 2 binding phosphatidylinositol 3-kinase binding protein phosphatase 2A binding phosphatidylinositol-4-phosphate binding 14-3-3 protein binding cadherin binding involved in cell-cell adhesion protein N-terminus binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter MAPK cascade activation of MAPKKK activity angiogenesis negative regulation of protein phosphorylation inflammatory response activation of signaling protein activity involved in unfolded protein response cell cycle I-kappaB phosphorylation activation of JUN kinase activity negative regulation of cell proliferation extrinsic apoptotic signaling pathway via death domain receptors negative regulation of endothelial cell migration negative regulation of epithelial cell migration negative regulation of epithelial to mesenchymal transition positive regulation of neuron projection development negative regulation of phosphatidylinositol 3-kinase signaling negative regulation of angiogenesis cell motility involved in cerebral cortex radial glia guided migration layer formation in cerebral cortex negative regulation of vascular endothelial growth factor receptor signaling pathway negative regulation of NF-kappaB transcription factor activity negative regulation of toll-like receptor 4 signaling pathway negative regulation of GTPase activity tube formation negative regulation of catenin import into nucleus cellular response to vascular endothelial growth factor stimulus vascular endothelial growth factor receptor-2 signaling pathway reelin-mediated signaling pathway regulation of growth negative regulation of epidermal growth factor receptor signaling pathway positive regulation of apoptotic process regulation of GTPase activity regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of I-kappaB kinase/NF-kappaB signaling regulation of protein complex assembly negative regulation of MAP kinase activity positive regulation of MAPK cascade regulation of protein heterodimerization activity positive regulation of JUN kinase activity positive regulation of GTPase activity negative regulation of phosphatidylinositol 3-kinase activity cellular protein catabolic process innate immune response positive regulation of protein catabolic process negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of JNK cascade negative regulation of Ras protein signal transduction negative regulation of fibroblast proliferation neuron projection morphogenesis negative regulation of epithelial cell proliferation intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress negative regulation of G0 to G1 transition negative regulation of ERK1 and ERK2 cascade positive regulation of cell cycle arrest cellular response to lipopolysaccharide cellular response to interleukin-1 cellular response to tumor necrosis factor cellular response to epidermal growth factor stimulus negative regulation of protein serine/threonine kinase activity positive regulation of protein serine/threonine kinase activity endothelial cell apoptotic process negative regulation of canonical Wnt signaling pathway positive regulation of synapse maturation cell-cell adhesion positive regulation of dendrite development regulation of p38MAPK cascade negative regulation of vascular endothelial growth factor signaling pathway positive regulation of proteasomal protein catabolic process positive regulation of IRE1-mediated unfolded protein response negative regulation of cyclin catabolic process positive regulation of neuron migration positive regulation of apoptotic signaling pathway immune system process apoptotic process response to unfolded protein signal transduction multicellular organism development negative regulation of cell growth
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0