Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • DACT1 Antibodies

          Invitrogen

          DACT1 Polyclonal Antibody

          View all (9) DACT1 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite DACT1 Polyclonal Antibody

          • Antibody Testing Data (1)
          DACT1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DACT1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DACT1 Antibody (PA5-114208) in WB

          Western Blot of Human 293T using a DACT1 Polyclonal Antibody (Product # PA5-114208). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DACT1 Antibody in Western Blot (WB)

          Product Details

          PA5-114208

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1.0 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Immunogen sequence: LSLVQKKTHP VRTNKPRTSV NADPTKGLLR NGSVCVRAPG GVSQGNSVNL
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS with 2% sucrose

          Contains

          0.09% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          RRID

          AB_2884722

          Product Specific Information

          Immunogen sequence: LSLVQKKTHP VRTNKPRTSV NADPTKGLLR NGSVCVRAPG GVSQGNSVNL

          For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.

          Predicted homology: Cow: 100%; Dog: 93%; Guinea Pig: 92%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 92%; Rat: 100%; Zebrafish: 85%

          Target Information

          Dapper homolog 1, an ortholog of Xenopus Dpr, antagonizes Wnt signaling by promoting Dvl degradation via a lysosome inhibitor-sensitive and proteasome inhibitor-insensitive mechanism. Dapper homolog 1 acts as an interacting protein for Dvl, and the interaction is formed between the DEP (Dishevelled, Egl-10, and pleckstrin) domain of Dvl and the central and C-terminal regions of Dapper homolog 1. The gene for human Dapper homolog 1 is localized on chromosomal region 14q23.1. The protein impedes the degradation of CTNNB1/beta-catenin, thereby enhancing the transcriptional activation of target genes of the Wnt signaling pathway. The protein is also required for normal notochord formation and is downregulated in hepatocellular carcinomas (HCC). It is expressed in human liver and various other tissues.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Dapper antagonist of catenin 1; Dapper homolog 1; dapper, antagonist of beta-catenin, homolog 1; dishevelled-binding antagonist of beta-catenin 1; hDPR1; Hepatocellular carcinoma novel gene 3 protein; heptacellular carcinoma novel gene 3

          View more View less

          Gene Aliases: DACT1; DAPPER; DAPPER1; DPR1; FRODO; HDPR1; HNG3; THYEX3

          View more View less

          UniProt ID: (Human) Q9NYF0

          View more View less

          Entrez Gene ID: (Human) 51339

          View more View less

          Function(s)
          RNA polymerase II transcription factor binding protein kinase C binding protein binding beta-catenin binding histone deacetylase binding protein kinase A binding delta-catenin binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter Wnt signaling pathway neural tube development positive regulation of Wnt signaling pathway negative regulation of Wnt signaling pathway regulation of protein stability negative regulation of protein binding positive regulation of protein binding regulation of catenin import into nucleus positive regulation of catenin import into nucleus positive regulation of protein catabolic process negative regulation of JNK cascade embryonic hindgut morphogenesis negative regulation of canonical Wnt signaling pathway regulation of nodal signaling pathway positive regulation of cellular protein catabolic process negative regulation of beta-catenin-TCF complex assembly regulation of Wnt signaling pathway, planar cell polarity pathway negative regulation of G1/S transition of mitotic cell cycle
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-48zfn:80/100.66.135.186:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0