Product References
Zinc Binding to S100B Affords Regulation of Trace Metal Homeostasis and Excitotoxicity in the Brain.
Frontiers in molecular neuroscience
Hagmeyer S,Cristóvão JS,Mulvihill JJE,Boeckers TM,Gomes CM,Grabrucker AM
Wed Sep 30 00:00:00 UTC 2020
Dual T cell- and B cell-intrinsic deficiency in humans with biallelic RLTPR mutations.
The Journal of experimental medicine
Wang Y,Ma CS,Ling Y,Bousfiha A,Camcioglu Y,Jacquot S,Payne K,Crestani E,Roncagalli R,Belkadi A,Kerner G,Lorenzo L,Deswarte C,Chrabieh M,Patin E,Vincent QB,Müller-Fleckenstein I,Fleckenstein B,Ailal F,Quintana-Murci L,Fraitag S,Alyanakian MA,Leruez-Ville M,Picard C,Puel A,Bustamante J,Boisson-Dupuis S,Malissen M,Malissen B,Abel L,Hovnanian A,Notarangelo LD,Jouanguy E,Tangye SG,Béziat V,Casanova JL
Mon Oct 17 00:00:00 UTC 2016
Impaired RASGRF1/ERK-mediated GM-CSF response characterizes CARD9 deficiency in French-Canadians.
The Journal of allergy and clinical immunology
Gavino C,Hamel N,Zeng JB,Legault C,Guiot MC,Chankowsky J,Lejtenyi D,Lemire M,Alarie I,Dufresne S,Boursiquot JN,McIntosh F,Langelier M,Behr MA,Sheppard DC,Foulkes WD,Vinh DC
Fri Apr 01 00:00:00 UTC 2016
Proline isomerisation as a novel regulatory mechanism for p38MAPK activation and functions.
Cell death and differentiation
Brichkina A,Nguyen NT,Baskar R,Wee S,Gunaratne J,Robinson RC,Bulavin DV
Sat Oct 01 00:00:00 UTC 2016
M10, a caspase cleavage product of the hepatocyte growth factor receptor, interacts with Smad2 and demonstrates antifibrotic properties in vitro and in vivo.
Translational research : the journal of laboratory and clinical medicine
Atanelishvili I,Shirai Y,Akter T,Buckner T,Noguchi A,Silver RM,Bogatkevich GS
Fri Apr 01 00:00:00 UTC 2016
Emodin isolated from Polygoni Multiflori Ramulus inhibits melanogenesis through the liver X receptor-mediated pathway.
Chemico-biological interactions
Kim MO,Park YS,Nho YH,Yun SK,Kim Y,Jung E,Paik JK,Kim M,Cho IH,Lee J
Mon Apr 25 00:00:00 UTC 2016
CDK1 phosphorylates WRN at collapsed replication forks.
Nature communications
Palermo V,Rinalducci S,Sanchez M,Grillini F,Sommers JA,Brosh RM,Zolla L,Franchitto A,Pichierri P
Fri Sep 16 00:00:00 UTC 2016
Golgi self-correction generates bioequivalent glycans to preserve cellular homeostasis.
eLife
Mkhikian H,Mortales CL,Zhou RW,Khachikyan K,Wu G,Haslam SM,Kavarian P,Dell A,Demetriou M
Wed Jun 08 00:00:00 UTC 2016
MicroRNA-602 and microRNA-608 regulate sonic hedgehog expression via target sites in the coding region in human chondrocytes.
Arthritis & rheumatology (Hoboken, N.J.)
Akhtar N,Makki MS,Haqqi TM
Sun Feb 01 00:00:00 UTC 2015
Germline Heterozygous Variants in SEC23B Are Associated with Cowden Syndrome and Enriched in Apparently Sporadic Thyroid Cancer.
American journal of human genetics
Yehia L,Niazi F,Ni Y,Ngeow J,Sankunny M,Liu Z,Wei W,Mester JL,Keri RA,Zhang B,Eng C
Thu Nov 05 00:00:00 UTC 2015
Label-free detection microarray for novel peptide ligands screening base on MS-SPRi combination.
Talanta
Wang W,Zhang D,Wei Z,Wang Z,Bu X,Yang S,Fang Q,Hu Z
Sun Mar 01 00:00:00 UTC 2015
RhoA Kinase (Rock) and p90 Ribosomal S6 Kinase (p90Rsk) phosphorylation of the sodium hydrogen exchanger (NHE1) is required for lysophosphatidic acid-induced transport, cytoskeletal organization and migration.
Cellular signalling
Wallert MA,Hammes D,Nguyen T,Kiefer L,Berthelsen N,Kern A,Anderson-Tiege K,Shabb JB,Muhonen WW,Grove BD,Provost JJ
Sun Mar 01 00:00:00 UTC 2015
SIRT6 deficiency culminates in low-turnover osteopenia.
Bone
Sugatani T,Agapova O,Malluche HH,Hruska KA
Tue Dec 01 00:00:00 UTC 2015
Infectious bursal disease virus VP5 polypeptide: a phosphoinositide-binding protein required for efficient cell-to-cell virus dissemination.
PloS one
Méndez F,de Garay T,Rodríguez D,Rodríguez JF
Fri Jan 08 00:00:00 UTC 2016
The nuclear receptor nr4a1 controls CD8 T cell development through transcriptional suppression of runx3.
Scientific reports
Nowyhed HN,Huynh TR,Blatchley A,Wu R,Thomas GD,Hedrick CC
Thu Mar 12 00:00:00 UTC 2015
Methionine Mistranslation Bypasses the Restraint of the Genetic Code to Generate Mutant Proteins with Distinct Activities.
PLoS genetics
Wang X,Pan T
Tue Dec 01 00:00:00 UTC 2015
The palmitoyl acyltransferase HIP14 shares a high proportion of interactors with huntingtin: implications for a role in the pathogenesis of Huntington's disease.
Human molecular genetics
Butland SL,Sanders SS,Schmidt ME,Riechers SP,Lin DT,Martin DD,Vaid K,Graham RK,Singaraja RR,Wanker EE,Conibear E,Hayden MR
Fri Aug 01 00:00:00 UTC 2014
ATM-mediated Mad1 Serine 214 phosphorylation regulates Mad1 dimerization and the spindle assembly checkpoint.
Carcinogenesis
Yang C,Hao J,Kong D,Cui X,Zhang W,Wang H,Guo X,Ma S,Liu X,Pu P,Xu B
Mon Sep 01 00:00:00 UTC 2014
Ligand binding alters dimerization and sequestering of urokinase receptors in raft-mimicking lipid mixtures.
Biophysical journal
Ge Y,Siegel AP,Jordan R,Naumann CA
Tue Nov 04 00:00:00 UTC 2014
The RNA-binding protein, ZFP36L2, influences ovulation and oocyte maturation.
PloS one
Ball CB,Rodriguez KF,Stumpo DJ,Ribeiro-Neto F,Korach KS,Blackshear PJ,Birnbaumer L,Ramos SB
Thu Jan 22 00:00:00 UTC 2015
Modulation of TET2 expression and 5-methylcytosine oxidation by the CXXC domain protein IDAX.
Nature
Ko M,An J,Bandukwala HS,Chavez L,Aijö T,Pastor WA,Segal MF,Li H,Koh KP,Lähdesmäki H,Hogan PG,Aravind L,Rao A
Thu May 02 00:00:00 UTC 2013
Exon B of human surfactant protein A2 mRNA, alone or within its surrounding sequences, interacts with 14-3-3; role of cis-elements and secondary structure.
American journal of physiology. Lung cellular and molecular physiology
Noutsios GT,Silveyra P,Bhatti F,Floros J
Sat Jun 01 00:00:00 UTC 2013
Hrk mediates 2-methoxyestradiol-induced mitochondrial apoptotic signaling in prostate cancer cells.
Molecular cancer therapeutics
Chang I,Majid S,Saini S,Zaman MS,Yamamura S,Chiyomaru T,Shahryari V,Fukuhara S,Deng G,Dahiya R,Tanaka Y
Sat Jun 01 00:00:00 UTC 2013
Missense mutations in the NF2 gene result in the quantitative loss of merlin protein and minimally affect protein intrinsic function.
Proceedings of the National Academy of Sciences of the United States of America
Yang C,Asthagiri AR,Iyer RR,Lu J,Xu DS,Ksendzovsky A,Brady RO,Zhuang Z,Lonser RR
Tue Mar 22 00:00:00 UTC 2011
Superoxide dismutase 1 (SOD1) is a target for a small molecule identified in a screen for inhibitors of the growth of lung adenocarcinoma cell lines.
Proceedings of the National Academy of Sciences of the United States of America
Somwar R,Erdjument-Bromage H,Larsson E,Shum D,Lockwood WW,Yang G,Sander C,Ouerfelli O,Tempst PJ,Djaballah H,Varmus HE
Tue Sep 27 00:00:00 UTC 2011
Polypyrimidine tract-binding protein is critical for the turnover and subcellular distribution of CD40 ligand mRNA in CD4+ T cells.
Journal of immunology (Baltimore, Md. : 1950)
Matus-Nicodemos R,Vavassori S,Castro-Faix M,Valentin-Acevedo A,Singh K,Marcelli V,Covey LR
Tue Feb 15 00:00:00 UTC 2011
Heparan sulfate modification of the transmembrane receptor CD47 is necessary for inhibition of T cell receptor signaling by thrombospondin-1.
The Journal of biological chemistry
Kaur S,Kuznetsova SA,Pendrak ML,Sipes JM,Romeo MJ,Li Z,Zhang L,Roberts DD
Fri Apr 29 00:00:00 UTC 2011
[Changes in the urinary excretion of gamma-glutamyltranspeptidase, leucine aminopeptidas and alkaline phosphatase in the combined action of ethylene glycol and high temperature].
Problemi na khigienata
Koleva M
Fri Sep 01 00:00:00 UTC 1978