Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Gemin 3 Antibodies

          Abnova

          DDX20 Polyclonal Antibody, MaxPab™

          View all (18) Gemin 3 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite DDX20 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          DDX20 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DDX20 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DDX20 Antibody (H00011218-B01P) in WB

          Western Blot analysis of DDX20 expression in transfected 293T cell line by DDX20 MaxPab polyclonal antibody. Lane 1: DDX20 transfected lysate(90.64 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DDX20 Antibody in Western Blot (WB)

          Product Details

          H00011218-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          DDX20 (AAH34953.1, 1 a.a. approximately 824 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAAAFEASGA LAAVATAMPA EHVAVQVPAP EPTPGPVRIL RTAQDLSSPR TRTGDVLLAE PADFESLLLS RPVLEGLRAA GFERPSPVQL KAIPLGRCGL DLIVQAKSGT GKTCVFSTIA LDSLVLENLS TQILILAPTR EIAVQIHSVI TAIGIKMEGL ECHVFIGGTP LSQDKTRLKK CHIAVGSPGR IKQLIELDYL NPGSIRLFIL DEADKLLEEG SFQEQINWIY SSLPASKQML AVSATYPEFL ANALTKYMRD PTFVRLNSSD PSLIGLKQYY KVVNSYPLAH KVFEEKTQHL QELFSRIPFN QALVFSNLHS RAQHLADILS SKGFPAECIS GNMNQNQRLD AMAKLKHFHC RVLISTDLTS RGIDAEKVNL VVNLDVPLDW ETYMHRIGRA GRFGTLGLTV TYCCRGEEGN MMMRIAQKCN INLLPLPDPI PSGLMEECVD WDVEVKAAVH TYGIASVPNQ PLKKQIQKIE RTLQIQKAHG DHMASSRNNS VSGLSVKSKN NTKQKLPVKS HSECGIIEKA TSPKELGCDR QSEEQMKNSV QTPVENSTNS QHQVKEALPV SLPQIPCLSS FKIHQPYTLT FAELVEDYEH YIKEGLEKPV EIIRHYTGPG DQTVNPQNGF VRNKVTEQRV PVLASSSQSG DSESDSDSYS SRTSSQSKGN KSYLEGSSDN QLKDSESTPV DDRISLEQPP NGSDTPNPEK YQESPGIQMK TRLKEGASQR AKQSRRNLPR RSSFRLQTEA QEDDWYDCHR EIRLSFSDTY QDYEEYWRAY YRAWQEYYAA ASHSYYWNAQ RHPSWMAAYH MNTIYLQEMM HSNQ

          Target Information

          Gemin 3 is a member of the DEAD box protein family which are characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD) and are putative RNA helicases. DEAD box proteins such as Gemin 3 are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of DEAD box family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. Gemin 3 has an ATPase activity and a component of the survival of motor neurons (SMN) complex. The Gemin 3 protein may play a catalytic role in the function of the SMN complex on RNPs. Diseases associated with Gemin 3 protein dysfunction include spinal muscular dystrophy and muscular atrophy.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Component of gems 3; DDX 20; DEAD (Asp-Glu-Ala-Asp) box polypeptide 20; DEAD box protein 20; DEAD box protein DP 103; DEAD-box protein DP103; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 20, 103kD; DEAD/H BOX 20; DKFZP434H052; DP 103; Gemin-3; Probable ATP-dependent RNA helicase DDX20; SMN-interacting protein

          View more View less

          Gene Aliases: DDX20; DP103; GEMIN3

          View more View less

          UniProt ID: (Human) Q9UHI6

          View more View less

          Entrez Gene ID: (Human) 11218

          View more View less

          Function(s)
          DNA binding ATP-dependent RNA helicase activity protein binding ATP binding protein domain specific binding protein binding, bridging histone deacetylase binding repressing transcription factor binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter spliceosomal tri-snRNP complex assembly spliceosomal snRNP assembly RNA processing negative regulation of cell proliferation RNA secondary structure unwinding positive regulation of apoptotic process oogenesis regulation of steroid biosynthetic process nuclear import
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-f7m9l:80/100.66.129.252:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0