Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
DEF6, or IBP, is a guanine nucleotide exchange factor for RAC and CDC42 that is highly expressed in B and T cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DEF-6; DEF6 guanine nucleotide exchange factor; Differentially expressed in FDCP 6; Differentially expressed in FDCP 6 homolog; IRF-4-binding protein; IRF4-binding protein; OTTHUMP00000016251; RP3-329A5.5; SWAP-70-like adapter of T-cells; SWAP-70-like adaptor protein of T cells
Gene Aliases: 2410003F05Rik; 6430538D02Rik; AV094905; DEF6; IBP; SLAT; Slat2; Slat6; SWAP70L
UniProt ID: (Human) Q9H4E7, (Mouse) Q8C2K1
Entrez Gene ID: (Human) 50619, (Mouse) 23853
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support