Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
May be involved in transcriptional regulation as a transcriptional corepressor. The DEPDC1A-ZNF224 complex may play a critical role in bladder carcinogenesis by repressing the transcription of the A20 gene, leading to transport of NF-KB protein into the nucleus, resulting in suppression of apoptosis of bladder cancer cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cell cycle control protein SDP35; DEP domain-containing protein 1A
Gene Aliases: DEP.8; DEPDC1; DEPDC1-V2; DEPDC1A; SDP35
UniProt ID: (Human) Q5TB30
Entrez Gene ID: (Human) 55635
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support