Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 6
Predicted band size: 46 kDa
Positive Control: MCF-7 cell lysate, Daudi cell lysate, PC-3M cell lysate, N2A cell lysate, L6 cell lysate, mouse brain tissue lysate, human small intestine tissue.
Sequence Similarities: 96.35%Mouse; 97.73% Rat.
Tissue Specificity: Expression is negatively regulated by both mTORC1 and mTORC2 (at protein level).
Negative regulator of the mTORC1 and mTORC2 signaling pathways. Inhibits the kinase activity of both complexes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DEP domain containing 6; DEP domain containing mTOR interacting protein; DEP domain containing MTOR-interacting protein; DEP domain-containing mTOR-interacting protein; DEP domain-containing protein 6; hDEPTOR
Gene Aliases: DEP.6; DEPDC6; DEPTOR; Kiaa4200; RGD1561030
UniProt ID: (Human) Q8TB45, (Mouse) Q570Y9
Entrez Gene ID: (Human) 64798, (Mouse) 97998, (Rat) 314979
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support