Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
DGCR6 may play a role in neural crest cell migration into the third and fourth pharyngeal pouches.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DiGeorge syndrome chromosome region 6; DiGeorge syndrome critical region 6; DiGeorge syndrome critical region 6 homolog; DiGeorge syndrome critical region protein 6; Protein DGCR6
Gene Aliases: DGCR6
UniProt ID: (Human) Q14129, (Mouse) O35347
Entrez Gene ID: (Human) 8214, (Mouse) 13353, (Rat) 303794
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support