Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MLFWVLGLLI LCGFLWTRKG KLKIEDITDK YIFITGCDSG FGNLAARTFD KKGFHVIAAC LTESGSTALK AETSERLRTV LLDVTDPENV KRTAQWVKNQ VGEKGLWGLI NNAGVPGVLA PTDWLTLEDY REPIEVNLFG LISVTLNMLP LVKKAQGRVI NVSSVGGRLA IVGGGYTPSK YAVEGFNDSL RRDMKAFGVH VSCIEPGLFK TNLADPVKVI EKKLAIWEQL SPDIKQQYGE GYIEKSLDKL KGNKSYVNMD LSPVVECMDH ALTSLFPKTH YAAGKDAKIF WIPLSHMPAA LQDFLLLKQK AELANPKAV
DHRS9 is a 3-alpha-hydroxysteroid dehydrogenase that converts 3-alpha-tetrahydroprogesterone (allopregnanolone) to dihydroxyprogesterone and 3-alpha-androstanediol to dihydroxyprogesterone.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-alpha hydroxysteroid dehydrogenase; 3-alpha-HSD; dehydrogenase/reductase (SDR family) member 9; Dehydrogenase/reductase SDR family member 9; NADP-dependent retinol dehydrogenase/reductase; RDH-E2; RDH-TBE; RDHL; Retinol dehydrogenase 15; retinol dehydrogenase homolog; Short chain dehydrogenase/reductase family 9C member 4; short chain dehydrogenase/reductase family 9C, member 4; Short-chain dehydrogenase/reductase retSDR8; Tracheobronchial epithelial cell-specific retinol dehydrogenase
Gene Aliases: 3-alpha-HSD; 3ALPHA-HSD; DHRS9; RDH-E2; RDH-TBE; RDH15; RDHL; RDHTBE; RETSDR8; SDR9C4; UNQ835/PRO1773
UniProt ID: (Human) Q9NRA9
Entrez Gene ID: (Human) 10170
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support