Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
DIRAS2 belongs to a distinct branch of the functionally diverse Ras superfamily of monomeric GTPases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DIRAS family GTP binding RAS like 2; DIRAS family, GTP-binding RAS-like 2; Distinct subgroup of the Ras family member 2; GTP-binding protein Di-Ras2
Gene Aliases: 2900052J15Rik; AI414999; Di-Ras2; DIRAS2
UniProt ID: (Human) Q96HU8, (Mouse) Q5PR73
Entrez Gene ID: (Human) 54769, (Mouse) 68203, (Rat) 291006
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support