Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEEDTDYRIR FSSLCFFNDH VGFHGTIKSS PSDFIVIEID EQGQLVNKTI DEPIFKISEI QLEPNNFPKK PKLDLQNLSL EDGRNQEVHT LIKYTDGDQN HQSGSEKEDT IVDGTSKCEE KADVLSSFLD EKTHELLNNF ACDVREKWLS KTELIGLPPE FSIGRIVDKN QRASLHSAIR QKFPFLVTVG KNSEIVVKPN LEYKELCHLV SEEEAFDFFK YLDAKKENSK FTFKPDTNKD HRKAVHHFVN KKFGNLVETK SFSKMNCSAG NPNVVVTVRF REKAHKRGKR PLSECQEGKV IYTAFTLRKE NLEMFEAIGF LAIKLGVIPS DFSYAGLKDK KAITYQAMVV RKVTPERLKN IEKEIEKKRM NVFNIRSVDD SLRLGQLKGN HFDIVIRNLK KQINDSANLR ERIMEAIENV KKKGFVNYYG PQRFGKGRKV HTDQIGLALL KNEMMKAIKL FLTPEDLDDP VNRAKKYFLQ TEDAKGTLSL MPEFKVRERA LLEALHRFGM TEEGCIQAWF SLPHSMRIFY VHAYTSKIWN EAVSYRLETY GARVVQGDLV CLDEDIDDEN FQNSKIHLVT EEEGSANMYA IHQVVLPVLG YNIQYPKNKV GQWYHDILSR DGLQTCRFKV PTLKLNIPGC YRQILKHPCN LSYQLMEDHD IDVKTKGSHI DETALSLLIS FDLDASCYAT VCLKEIMKHD V
Pseudouridylate synthase that catalyzes pseudouridylation of RNAs. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: pseudouridylate synthase 7 homolog (S. cerevisiae)-like; Pseudouridylate synthase 7 homolog-like protein; pseudouridylate synthase 7-like; Pseudouridylate synthase PUS7L
Gene Aliases: PUS7L
UniProt ID: (Human) Q9H0K6
Entrez Gene ID: (Human) 83448
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support