Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: ALSYPQEEAT LNEMFREVEE LMEDTQHKLR SAVEEMEAEE AAAKASSEVN LANLPPSYHN ETNTDTKVGN NTIHVHREIH KITNNQTGQM VFSETVITSV GDEEGRRSHE CIIDEDCGPS MYCQFASFQY TCQPCRGQRM LCTRDSECCG DQLCVWGHCT KMATRGSNGT ICDNQRDCQP GLCCAFQRGL LFPVCTPLPV EGELCHDPAS RLLDLITWEL EPDGALDRCP CASGLLCQPH SHSLVYVCKP TFVGSRDQDG EILLPREVPD EYEVGSFMEE VRQELEDLER SLTEEMALGE PAAAAAALLG GEEI
DKK3, like DKK1, DKK2, and DKK4, possesses an N-terminal signal peptide and 2 conserved cysteine-rich domains, which are separated by a linker region and contain 10 cysteine residues each. The second cysteine region has a putative lipid-binding function that may facilitate WNT/DKK interactions at the plasma membrane. The linker region contains 50 to 55 amino acids in DKK1, DKK2, and DKK4, whereas in DKK3 it contains only 12 amino acids. All DKKs have several potential sites for cleavage by furin-type proteases. Northern blot analysis revealed wide expression of the DKK3 transcript, with highest expression in heart, brain, and spinal cord. In situ hybridization reveals highest expression in mouse brain, eye, and heart.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dickkopf; dickkopf 3; dickkopf 3 homolog; Dickkopf gene 3; dickkopf homolog 3; Dickkopf-3; Dickkopf-related protein 3; Dickkopf3; DKK 3; dkk-3; hDkk 3; hDkk-3; hhDkk3; mDkk-3; regulated in glioma; RIG-like 5-6; RIG-like 7-1; Xenopus
Gene Aliases: AW061014; C87148; DKK3; REIC; RIG; UNQ258/PRO295
UniProt ID: (Human) Q9UBP4, (Mouse) Q9QUN9
Entrez Gene ID: (Human) 27122, (Mouse) 50781
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support